Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Immunogen sequence: MSGDHLHNDS QIEADFRLND SHKHKDKHKD REHRHKEHKK EKDREKSKHS NSEHKDSEKK HKEKEKTKHK DGSSEKHKDK HKDRDKEKRK EEKVRASGDA
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; type I DNA topoisomerase
Gene Aliases: AI467334; D130064I21Rik; Top-1; TOP1; TOPI
UniProt ID: (Human) P11387, (Mouse) Q04750
Entrez Gene ID: (Human) 7150, (Mouse) 21969
Molecular Function: DNA topoisomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support