Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • TPM1 Antibodies

          Invitrogen

          TPM1 Monoclonal Antibody (TM31)

          View all (16) TPM1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TPM1 Monoclonal Antibody (TM31)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TPM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TPM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TPM1 Antibody (MA5-32982) in WB

          Western blot analysis of Tropomyosin(36/39 kDa) in, Lane 1: rat skeletal muscle tissue lysates, Lane 2: rat heart tissue lysates, Lane 3: mouse skeletal muscle tissue lysates, Lane 4: mouse heart tissue lysates. Electrophoresis was performed on a 5-20% SDS-PAGE gel at 70V (Stacking gel) / 90V (Resolving gel) for 2-3 hours. The sample well of each lane was loaded with 30 µg of sample under reducing conditions. After Electrophoresis, proteins were transferred to a nitrocellulose membrane at 150 mA for 50-90 minutes. The membrane was blocked ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TPM1 Antibody in Western Blot (WB)
          TPM1 Monoclonal Antibody (TM31)

          Product Details

          MA5-32982

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          TM31

          Immunogen

          Chicken gizzard tropomyosin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          100 µg/mL

          Amount

          100 µg

          Storage buffer

          ascites/1.2% sodium acetate with 2mg BSA

          Contains

          0.01mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2802616

          Product Specific Information

          Reconstitute by adding 1 mL of PBS buffer, yielding a concentration of 100 µg/mL.

          Positive Control - WB: rat skeletal muscle tissue, rat heart tissue, mouse skeletal muscle tissue, mouse heart tissue.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

          Target Information

          Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha tropomyosin; alpha tropomyosin of brain; alpha tropomyosin of fibroblasts F2; alpha-Tm-1; alpha-Tm-2 isoform; Alpha-tropomyosin; alpha-tropomyosin (partial); alpha-tropomyosin 1; alpha-tropomyosin 2; alpha-tropomyosin of skeletal fast muscle; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; hepatoma alpha tropomyosin; myofibrillar protein; component of thin filaments; alternatively spliced; sarcomeric tropomyosin kappa; skeletal muscle tropomyosin; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; TPM1-alpha; tropomyosin; tropomyosin (CTm4); tropomyosin (CTm7); tropomyosin 1 (alpha); tropomyosin 1 (alpha) isoform TPM1_1; tropomyosin 1 (alpha) isoform TPM1_2; tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin-1; unnamed protein product

          View more View less

          Gene Aliases: AA986836; AI854628; alpha-fTM; Alpha-tm; C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; TM2; Tm3; Tma2; Tmpa; TMSA; Tpm-1; TPM1; Tpma; tropomyosin-1

          View more View less

          UniProt ID: (Chicken) P04268, (Human) P09493, (Rat) P04692, (Mouse) P58771

          View more View less

          Entrez Gene ID: (Chicken) 396366, (Human) 7168, (Rat) 24851, (Mouse) 22003

          View more View less

          Function(s)
          protein binding cytoskeletal protein binding identical protein binding protein homodimerization activity protein heterodimerization activity actin filament binding actin binding structural constituent of cytoskeleton structural constituent of muscle protein N-terminus binding actin binding motor protein
          Process(es)
          muscle contraction actin filament organization cardiac muscle contraction positive regulation of heart rate by epinephrine regulation of muscle contraction cytoskeleton organization regulation of heart contraction regulation of cell shape muscle filament sliding negative regulation of cell migration ruffle organization cellular response to reactive oxygen species wound healing sarcomere organization positive regulation of cell adhesion positive regulation of stress fiber assembly ventricular cardiac muscle tissue morphogenesis negative regulation of vascular smooth muscle cell proliferation negative regulation of vascular associated smooth muscle cell migration in utero embryonic development positive regulation of ATPase activity regulation of ATPase activity actin filament capping embryo development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0