Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TRAF3IP2 Antibodies

          Invitrogen

          TRAF3IP2 Polyclonal Antibody

          View all (14) TRAF3IP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TRAF3IP2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          TRAF3IP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRAF3IP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRAF3IP2 Antibody (PA5-110513) in ICC/IF

          Immunofluorescence analysis of TRAF3IP2 in NIH/3T3 cells. Samples were incubated with TRAF3IP2 Polyclonal antibody (Product # PA5-110513) using a dilution of 1:50 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRAF3IP2 Antibody in Immunocytochemistry (ICC/IF)
          TRAF3IP2 Antibody in Immunocytochemistry (ICC/IF)
          TRAF3IP2 Antibody in Immunocytochemistry (ICC/IF)
          TRAF3IP2 Antibody in Western Blot (WB)
          TRAF3IP2 Antibody in Western Blot (WB)
          TRAF3IP2 Polyclonal Antibody

          Product Details

          PA5-110513

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRAF3IP2 (NP_6717332)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.68 mg/mL

          Amount

          68 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2855924

          Product Specific Information

          Immunogen sequence: MPPQLQETRM NRSIPVEVDE SEPYPSQLLK PIPEYSPEEE SEPPAPNIRN MAPNSLSAPT MLHNSSGDFS QAHSTLKLAN HQRPVSRQVT CLRTQVLEDS EDSFCRRHPG LGKAFPSGCS AVSEPASESV VGALPAEHQF SFMEKRNQWL VSQLSAASPD TGHDSDKSDQ SLPNASADSL GGSQEMVQRP QPHRNRAGLD LPTIDTGYDS QPQDVLGIRQ LERPLPLTSV CYPQDLPRPL RSREFPQFEP QRYPACAQML PPNLSPHAPW NYHYHCPGSP DHQVPYGHDY PRAAYQQVIQ

          Target Information

          This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Two alternative transcripts encoding different proteins have been identified. A third transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ACT1; Adapter protein CIKS; Connection to IKK and SAPK/JNK; E3 ubiquitin ligase TRAF3IP2; E3 ubiquitin-protein ligase CIKS; NFkB-activating protein ACT1; Nuclear factor NF-kappa-B activator 1; ORF1; ORF2; ORF3; RP3-487J7.3; symbol withdrawn, see C6orf4; TRAF3-interacting protein 2; unnamed protein product

          View more View less

          Gene Aliases: ACT1; AI429613; C6orf2; C6orf4; C6orf5; C6orf6; CANDF8; CIKS; PSORS13; RGD1304768; TRAF3IP2

          View more View less

          UniProt ID: (Human) O43734, (Mouse) Q8N7N6

          View more View less

          Entrez Gene ID: (Human) 10758, (Mouse) 103213, (Rat) 361857

          View more View less

          Function(s)
          receptor binding protein binding transferase activity ubiquitin protein ligase activity scaffold/adaptor protein
          Process(es)
          protein polyubiquitination establishment of T cell polarity B cell homeostasis B cell apoptotic process kidney development positive regulation of defense response to virus by host leukocyte activation involved in inflammatory response transitional two stage B cell differentiation B cell affinity maturation eosinophil mediated immunity protein import into nucleus inflammatory response immune response humoral immune response heart development response to xenobiotic stimulus gene expression protein ubiquitination cytokine-mediated signaling pathway B cell mediated immunity signal transduction involved in regulation of gene expression CD40 signaling pathway T cell differentiation tumor necrosis factor-mediated signaling pathway response to cytokine protein localization to nucleus intracellular signal transduction interleukin-17A-mediated signaling pathway type 2 immune response T cell activation neutrophil activation positive regulation of I-kappaB kinase/NF-kappaB signaling skin development mRNA stabilization lymph node development spleen development mucus secretion protein K63-linked ubiquitination cellular response to cytokine stimulus T-helper 17 type immune response cellular response to interleukin-17 interleukin-17-mediated signaling pathway protein localization to P-body negative regulation of interleukin-17-mediated signaling pathway eosinophil homeostasis immunoglobulin secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline