Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TRAF6 Antibodies

          Invitrogen

          TRAF6 Polyclonal Antibody

          6 Published Figures
          4 References
          View all (27) TRAF6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TRAF6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          • Published Figures (6)
          TRAF6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRAF6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRAF6 Antibody (PA5-29622) in ICC/IF

          Immunocytochemistry-Immunofluorescence analysis of TRAF6 was performed in HeLa cells fixed in 4% paraformaldehyde at RT for 15 min. Green: TRAF6 Polyclonal Antibody (Product # PA5-29622) diluted at 1:100. Red: alpha Tubulin, a cytoskeleton marker. Blue: Hoechst 33342 staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRAF6 Antibody in Immunocytochemistry (ICC/IF)
          TRAF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Immunoprecipitation (IP)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)
          TRAF6 Antibody in Western Blot (WB)

          Product Details

          PA5-29622

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -

          Immunoprecipitation (IP)

          1:100-1:500
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human TRAF6. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          17 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 1% BSA, 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2547097

          Product Specific Information

          Recommended positive controls: A431, A549, HeLa, HepG2, NIH-3T3, BCL-1, PC-12, Rat2.

          Predicted reactivity: Mouse (80%), Pig (83%), Rhesus Monkey (97%), Bovine (82%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: E3 ubiquitin-protein ligase TRAF6; Interleukin-1 signal transducer; RING finger protein 85; RING-type E3 ubiquitin transferase TRAF6; TNF receptor-associated factor 6; TNF receptor-associated factor 6, E3 ubiquitin protein ligase; unnamed protein product

          View more View less

          Gene Aliases: 2310003F17Rik; AI851288; C630032O20Rik; MGC:3310; RNF85; TRAF6

          View more View less

          UniProt ID: (Human) Q9Y4K3, (Rat) B5DF45, (Mouse) P70196

          View more View less

          Entrez Gene ID: (Human) 7189, (Rat) 311245, (Mouse) 22034

          View more View less

          Function(s)
          ubiquitin-protein transferase activity tumor necrosis factor receptor binding protein binding zinc ion binding transferase activity enzyme binding protein binding, bridging ubiquitin conjugating enzyme binding ubiquitin-ubiquitin ligase activity signaling adaptor activity identical protein binding histone deacetylase binding protein kinase B binding metal ion binding ubiquitin protein ligase activity ligase activity protein kinase binding mitogen-activated protein kinase kinase kinase binding ubiquitin protein ligase binding thioesterase binding protein N-terminus binding scaffold/adaptor protein
          Process(es)
          autophagosome assembly negative regulation of transcription from RNA polymerase II promoter protein polyubiquitination ossification in utero embryonic development neural tube closure stimulatory C-type lectin receptor signaling pathway immune system process regulation of immunoglobulin production positive regulation of T cell cytokine production cytoplasmic pattern recognition receptor signaling pathway MyD88-dependent toll-like receptor signaling pathway immune response cellular response to DNA damage stimulus signal transduction I-kappaB kinase/NF-kappaB signaling mRNA transcription organ morphogenesis protein ubiquitination antigen processing and presentation of exogenous peptide antigen via MHC class II CD40 signaling pathway osteoclast differentiation positive regulation of protein ubiquitination lipopolysaccharide-mediated signaling pathway positive regulation of lipopolysaccharide-mediated signaling pathway activation of protein kinase activity positive regulation of type I interferon production positive regulation of interleukin-12 production positive regulation of interleukin-2 production positive regulation of interleukin-6 production tumor necrosis factor-mediated signaling pathway toll-like receptor 3 signaling pathway toll-like receptor 4 signaling pathway TRIF-dependent toll-like receptor signaling pathway NIK/NF-kappaB signaling Fc-epsilon receptor signaling pathway interleukin-33-mediated signaling pathway interleukin-17A-mediated signaling pathway T-helper 1 type immune response positive regulation of T cell proliferation odontogenesis of dentin-containing tooth defense response to bacterium regulation of apoptotic process myeloid dendritic cell differentiation regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of JUN kinase activity innate immune response bone resorption positive regulation of osteoclast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade bone remodeling cell development T cell receptor signaling pathway positive regulation of NF-kappaB transcription factor activity protein autoubiquitination interleukin-1-mediated signaling pathway protein K63-linked ubiquitination response to interleukin-1 protein K48-linked ubiquitination cellular response to lipopolysaccharide cellular response to cytokine stimulus interleukin-17-mediated signaling pathway regulation of neurotransmitter receptor localization to postsynaptic specialization membrane antiviral innate immune response intracellular signaling cassette protein branched polyubiquitination cellular response to oxygen-containing compound positive regulation of leukocyte adhesion to vascular endothelial cell protein complex assembly activation of NF-kappaB-inducing kinase activity JNK cascade cytokine-mediated signaling pathway positive regulation of interleukin-12 biosynthetic process positive regulation of interleukin-6 biosynthetic process positive regulation of smooth muscle cell proliferation regulation of immunoglobulin secretion positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of transcription regulatory region DNA binding
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-78576c459-fhm29:80/100.66.76.150:80.
          git-commit: 65c7ea78bfc366f1196f6925435a6e3b75c9e6fa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline