PA5-21317 targets TRAP in WB and IHC-P applications and shows reactivity with Human and mouse samples.
The PA5-21317 immunogen is a recombinant fragment corresponding to a region within amino acids 28 and 325 of TRAP (Uniprot ID#P13686).
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.