Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NIBP Antibodies

          Proteintech

          TRAPPC9 Polyclonal Antibody

          15 References
          View all (7) NIBP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TRAPPC9 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (11)
          TRAPPC9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRAPPC9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRAPPC9 Antibody (16014-1-AP) in ICC/IF

          Immunofluorescent analysis of HEK-293 cells using 16014-1-AP (TRAPPC9 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRAPPC9 Antibody in Immunocytochemistry (ICC/IF)
          TRAPPC9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAPPC9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAPPC9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAPPC9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAPPC9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRAPPC9 Antibody in Western Blot (WB)
          TRAPPC9 Antibody in Western Blot (WB)
          TRAPPC9 Antibody in Western Blot (WB)
          TRAPPC9 Antibody in Western Blot (WB)
          TRAPPC9 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TRAPPC9 Polyclonal Antibody

          Product Details

          16014-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TRAPPC9 Fusion Protein Ag8791 (563-912 aa encoded by BC006206)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.35 mg/mL

          Amount

          52.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: SCQENLLQD LSDDGISVSG FPLSSPFRQV VRPRVEGKPV NPPESNKAGD YSHVKTLEAV LNFKYSGGPG HTEGYYRNLS LGLHVEVEPS VFFTRVSTLP ATSTRQCHLL LDVFNSTEHE LTVSTRSSEA LILHAGECQR MAIQVDKFNF ESFPESPGEK GQFANPKQLE EERREARGLE IHSKLGICWR IPSLKRSGEA SVEGLLNQLV LEHLQLAPLQ WDVLVDGQPC DREAVAACQV GDPVRLEVRL TNRSPRSVGP FALTVVPFQD HQNGVHNYDL HDTVSFVGSS TFYLDAVQPS GQSACLGALL FLYTGDFFLH IRFHEDSTSK ELPPSWFCLP SVHVCALEAQ A (563-912 aa encoded by BC006206)

          Target Information

          NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: adaptor protein; IKK2 binding protein; NIK and IKK(beta) binding protein; NIK and IKK-beta binding protein; NIK and IKK{beta} binding protein; NIK- and IKBKB-binding protein; trafficking protein particle complex 9; TRAPP 120 kDa subunit; Tularik gene 1; tularik gene 1 protein; unnamed protein product

          View more View less

          Gene Aliases: 1810044A24Rik; 2900005P22Rik; 4632408O18Rik; IBP; IKBKBBP; mKIAA1882; MRT13; NIBP; RGD1309461; T1; TRS120; TRS130

          View more View less

          UniProt ID: (Human) Q96Q05, (Mouse) Q3U0M1

          View more View less

          Entrez Gene ID: (Human) 83696, (Rat) 315059, (Mouse) 76510

          View more View less

          Function(s)
          protein binding
          Process(es)
          cerebral cortex development neuron differentiation positive regulation of I-kappaB kinase/NF-kappaB signaling ER to Golgi vesicle-mediated transport vesicle coating
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-cvz79:80/100.66.78.224:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0