Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: FCRSCGLVL CEPCREADHQ PPGHCTLPVK EAAEERRRDF GEKLTRLREL MGELQRRKAA LEGVSKDLQA RYKAVLQEYG HEERRVQDEL ARSRKFFTGS LAEVEKSNSQ VVEEQSYLLN IAEVQAVSRC DYFLAKIKQA DVALLEETAD EEEPELTASL PRELTLQDVE LLKVGHVGPL QIGQAVKKPR TVNVEDSWAM EATASAASTS VTFREMDMSP EEVVASPRAS PAKQRGPEAA SNIQQCLFLK KMGAKGSTPG MFNLPVSLYV TSQGEVLVAD RGNYRIQVFT RKGFLKEIRR SPSGIDSFVL SFLGADLPNL T (111-430 aa encoded by BC003154)
TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 72 kDa Tat-interacting protein; bM468K2.2 (tripartite motif protein 32); RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; tripartite motif protein 32; tripartite motif-containing protein 32; unnamed protein product; zinc finger protein 117; zinc-finger protein HT2A
Gene Aliases: 1810045E12Rik; 3f3; BBS11; HT2A; LGMD2H; LGMDR8; TATIP; Zfp117
UniProt ID: (Human) Q13049, (Mouse) Q8CH72
Entrez Gene ID: (Human) 22954, (Rat) 313264, (Mouse) 69807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support