Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • TRPA1 Antibodies

          Invitrogen

          TRPA1 Polyclonal Antibody, DyLight™ 650

          View all (27) TRPA1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TRPA1 Polyclonal Antibody, DyLight™ 650

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TRPA1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          TRPA1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRPA1 Antibody (PA5-22832) in Flow

          Flow cytometry analysis of TRPA1 in A549 cells. Samples were incubated with TRPA1 polyclonal antibody (Product # PA5-22832) using a dilution of 2.5 µg/mL for 30 minutes at room temperature. An intracellular stain was performed with primary antibody (blue) and a matched isotype control (orange). Cells were fixed with 4% PFA and then permeabilized with 0.1% saponin. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRPA1 Antibody in Flow Cytometry (Flow)
          TRPA1 Polyclonal Antibody, DyLight™ 650

          Product Details

          PA5-22832

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:250
          -

          Immunohistochemistry (Frozen) (IHC (F))

          0.5-1.0 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          Assay-Dependent
          -

          ELISA (ELISA)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Guinea pig, Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein.
          3D Epitope / Immunogen

          Conjugate

          DyLight™ 650 DyLight™ 650 DyLight™ 650

          Excitation/Emission Max

          651/673 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.63 mg/mL

          Amount

          63 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          50mM sodium borate

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11152555

          Product Specific Information

          The target sequence has 69% sequence homology with rat.

          Suggested positive control: human brain membrane fraction.

          Target Information

          TRPA1 is a TRP-related channel that responds to cold temperatures and pungent compounds and plays a role in both nociceptor and hair cell transduction. TRPA1 is a transformation -associated gene product in lung epithelia, whereas its protein distribution is primarily restricted to sensory neurons. Blocking TRPA1 may be a therapeutic target for treating cold hyperalgesia caused by inflammation and nerve damage. It is now known that TRPA1 protein is also widely expressed outside of the CNS and is dys-regulated during oncogenic transformation. The structure of TRPA1 is highly related to both the protein ankyrin and transmembrane proteins. The specific function of TRPA1 has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ANKTM1; ankyrin-like with transmembrane domains 1; Ankyrin-like with transmembrane domains protein 1; ion channel; drTRPA1; nompC; p120; Transformation-sensitive protein p120; transient receptor potential A1; Transient receptor potential cation channel subfamily A member 1; TRPA-1; Wasabi receptor

          View more View less

          Gene Aliases: ANKTM1; FEPS; FEPS1; p120; TRPA1; TRPA1b

          View more View less

          UniProt ID: (Human) O75762, (Mouse) Q8BLA8, (Rat) Q6RI86

          View more View less

          Entrez Gene ID: (Human) 8989, (Mouse) 277328, (Guinea pig) 100526649, (Rat) 312896, (Zebrafish) 474351

          View more View less

          Function(s)
          ion channel activity voltage-gated calcium channel activity calcium channel activity channel activity calcium-release channel activity gated channel activity identical protein binding temperature-gated cation channel activity osmolarity-sensing cation channel activity protein binding mechanically-gated ion channel activity ligand-gated ion channel activity scaffold/adaptor protein
          Process(es)
          system process ion transport calcium ion transport cellular calcium ion homeostasis cell surface receptor signaling pathway response to cold response to xenobiotic stimulus urinary bladder smooth muscle contraction sensory perception of pain calcium-mediated signaling cellular response to stress ion transmembrane transport cellular response to heat positive regulation of insulin secretion involved in cellular response to glucose stimulus response to hydrogen peroxide response to pain thermoception detection of mechanical stimulus involved in sensory perception of pain detection of chemical stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception protein homotetramerization transmembrane transport cellular response to hydrogen peroxide cellular response to cold calcium ion transmembrane transport cellular response to food cellular response to carbon dioxide cellular response to caffeine cellular response to toxic substance calcium ion transmembrane import into cytosol regulation of neuronal action potential cellular response to oxygen-containing compound regulation of blood circulation positive regulation of monoatomic anion transport transport response to organic substance response to organic cyclic compound response to drug response to stimulus detection of chemical stimulus detection of temperature stimulus involved in thermoception response to fatty acid
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0