Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • TRPM8 Antibodies

          Proteintech

          TRPM8 Polyclonal Antibody

          5 References
          View all (32) TRPM8 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TRPM8 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          TRPM8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRPM8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRPM8 Antibody (12813-1-AP) in ICC/IF

          Immunofluorescent analysis of SH-SY5Y cells using 12813-1-AP (TRPM8 antibody) at dilution of 1:25 and Rhodamine-Goat anti-Rabbit IGG. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRPM8 Antibody in Immunocytochemistry (ICC/IF)
          TRPM8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPM8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPM8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPM8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPM8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPM8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPM8 Polyclonal Antibody

          Product Details

          12813-1-AP

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TRPM8 Fusion Protein Ag3483 (66-234 aa encoded by BC001135)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.15 mg/mL

          Amount

          22.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: SFLPVHTIV LIRENVCKCG YAQSQHMEGT QINQSEKWNY KKHTKEFPTD AFGDIQFETL GKKGKYIRLS CDTDAEILYE LLTQHWHLKT PNLVISVTGG AKNFALKPRM RKIFSRLIYI AQSKGAWILT GGTHYGLMKY IGEVVRDNTI SRSSEENIVA IGIAAWGMVS NRDTLIRNCD AEVPVGQEEV C (66-234 aa encoded by BC001135)

          Target Information

          TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: calcium channel; cation channel; Long transient receptor potential channel 6; LTrpC-6; MGC2849; short form of the TRPM8 cationic channel; sTRPM8-18; 18kDa non-channel protein; sM8alpha; sTRPM8-18; 18kDa non-channel protein; sTRPM8(2p-6b/+3pp); sTRPM8-26; 26kDa non-channel protein; sTRPM8(1-6b); sTRPM8-9; 9kDa non-channel protein; sM8beta; sTRPM8-9; 9kDa non-channel protein; sTRPM8(3p-6b); Transient receptor potential cation channel subfamily M member 8; Transient receptor potential p8; transient receptor potential subfamily M member 8; transmembrane ion channel; TRP-P8; Trp-p8; TRPM8 cationic channel; TRPM8 channel; TRPM8 isoform; 118kDa channel protein; TRPM8(3p-22b); TRPM8 isoform; 119kDa channel protein; TRPM8(3p-26.2); TRPM8 isoform; 121kDa channel protein; TRPM8(1-22p); TRPM8 isoform; 121kDa channel protein; TRPM8(2p-26.1); TRPM8 isoform; 127kDa channel protein; TRPM8(1-22b); TRPM8 isoform; 26kDa channel protein; TRPM8(18a-26.2); TRPM8 isoform; 27kDa channel protein; TRPM8(17p-26.1); TRPM8 isoform; 27kDa channel protein; TRPM8(17pp-26.2); TRPM8 isoform; 35kDa channel protein; TRPM8(17p-26.1); TRPM8 isoform; 35kDa channel protein; TRPM8(17pp-26.2); TRPM8 isoform; 76kDa channel protein; TRPM8(9p-22p); TRPM8 isoform; 83kDa channel protein; TRPM8(5a-26.2); TRPM8 isoform; 83kDa channel protein; TRPM8(9p-26.2); TRPM8 isoform; putative 106kDa channel protein; TRPM8(3p-26.2/+intron3p; TRPM8 isoform; putative 122kDa channel protein; TRPM8(3p-26.2/+intron3p); TRPM8 isoform; putative 17kDa non-channel protein; TRPM8(15a-26.2/-16b; TRPM8 isoform; putative 23kDa channel protein; TRPM8(16a-26.2/+intron16; TRPM8 isoform; putative 29kDa channel protein; TRPM8(17p-22p); TRPM8 isoform; putative 33kDa channel protein; TRPM8(18a-26.2/-23; TRPM8 isoform; putative 35kDa channel protein; TRPM8(17pp-22b); TRPM8 isoform; putative 37kDa channel protein; TRPM8(17pp-26.2/-25); TRPM8 isoform; putative 50kDa channel protein; TRPM8(16a-26.2/+intron16); TRPM8 isoform; putative 51kDa protein; TRPM8(2p-26.2/+4a; TRPM8 isoform; putative 73kDa channel protein; TRPM8(3p-26.2/+16d); TRPM8 isoform; putative 83kDa channel protein; TRPM8(3p-26.1/-7); TRPM8 isoform; putative 95kDa channel protein; TRPM8(3p-22b/-4b; unnamed protein product

          View more View less

          Gene Aliases: LTrpC-6; LTRPC6; trp-p8; TRPM8; TRPP8

          View more View less

          UniProt ID: (Human) Q7Z2W7

          View more View less

          Entrez Gene ID: (Human) 79054

          View more View less

          Function(s)
          ion channel activity calcium channel activity protein binding identical protein binding metal ion binding metal ion transmembrane transporter activity ligand-gated calcium channel activity
          Process(es)
          ion transport calcium ion transport cellular calcium ion homeostasis response to temperature stimulus response to cold calcium-mediated signaling ion transmembrane transport thermoception transmembrane transport calcium ion transmembrane transport positive regulation of cold-induced thermogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0