Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody specifically recognizes endogenous TTC7A.
Immunogen sequence: DATLKSKQD ELHRKALQTL ERAQQLAPSD PQVILYVSLQ LALVRQISSA MEQLQEALKV RKDDAHALHL LALLFSAQKH HQHALDVVNM AITEHPGNFN LMFTKVKLEQ VLKGPEEALV TCRQVLRLWQ TLYSFSQLGG LEKDGSFGEG LTMKKQSGMH LTLPDAHDAD SGSRRASSIA ASRLEEAMSE LTMPSSVLKQ GPMQLWTTLE QIWLQAAELF MEQQHLKEAG FCIQEAAGLF PTSHSVLYMR GRLAEVKGNL EEAKQLYKEA LTVNPDGVRI MHSLGLMLSR LGHKSLAQKV LRDAVERQST CHEAWQGL (149-465 aa encoded by BC114365)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TPR repeat protein 7A; unnamed protein product
Gene Aliases: 1110035E02Rik; 1700007L07Rik; fsn; GIDID; hea; MINAT; TTC7; Ttc7a
UniProt ID: (Human) Q9ULT0, (Mouse) Q8BGB2
Entrez Gene ID: (Human) 57217, (Mouse) 225049
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support