Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • GCP2 Antibodies

          Abnova

          TUBGCP2 Polyclonal Antibody, MaxPab™

          View all (5) GCP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TUBGCP2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TUBGCP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TUBGCP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TUBGCP2 Antibody (H00010844-D01P) in WB

          Western Blot analysis of TUBGCP2 expression in transfected 293T cell line by TUBGCP2 MaxPab polyclonal antibody. Lane 1: TUBGCP2 transfected lysate(102.50 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TUBGCP2 Antibody in Western Blot (WB)
          TUBGCP2 Polyclonal Antibody, MaxPab™

          Product Details

          H00010844-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TUBGCP2 (NP_006650.1, 1 a.a. approximately 902 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSEFRIHHDV NELLSLLRVH GGDGAEVYID LLQKNRTPYV TTTVSAHSAK VKIAEFSRTP EDFLKKYDEL KSKNTRNLDP LVYLLSKLTE DKETLQYLQQ NAKERAELAA AAVGSSTTSI NVPAAASKIS MQELEELRKQ LGSVATGSTL QQSLELKRKM LRDKQNKKNS GQHLPIFPAW VYERPALIGD FLIGAGISTD TALPIGTLPL ASQESAVVED LLYVLVGVDG RYVSAQPLAG RQSRTFLVDP NLDLSIRELV HRILPVAASY SAVTRFIEEK SSFEYGQVNH ALAAAMRTLV KEHLILVSQL EQLHRQGLLS LQKLWFYIQP AMRTMDILAS LATSVDKGEC LGGSTLSLLH DRSFSYTGDS QAQELCLYLT KAASAPYFEV LEKWIYRGII HDPYSEFMVE EHELRKERIQ EDYNDKYWDQ RYTIVQQQIP SFLQKMADKI LSTGKYLNVV RECGHDVTCP VAKEIIYTLK ERAYVEQIEK AFNYASKVLL DFLMEEKELV AHLRSIKRYF LMDQGDFFVH FMDLAEEELR KPVEDITPPR LEALLELALR MSTANTDPFK DDLKIDLMPH DLITQLLRVL AIETKQEKAM AHADPTELAL SGLEAFSFDY IVKWPLSLII NRKALTRYQM LFRHMFYCKH VERQLCSVWI SNKTAKQHSL HSAQWFAGAF TLRQRMLNFV QNIQYYMMFE VMEPTWHILE KNLKSASNID DVLGHHTGFL DTCLKDCMLT NPELLKVFSK LMSVCVMFTN CMQKFTQSMK LDGELGGQTL EHSTVLGLPA GAEERARKEL ARKHLAEHAD TVQLVSGFEA TINKFDKNFS AHLLDLLARL SIYSTSDCEH GMASVISRLD FNGFYTERLE RLSAERSQKA TPQVPVLRGP PAPAPRVAVT AQ

          Target Information

          The γ-Tubulin complex is composed of γ Tubulin and the γ-Tubulin complexassociated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. γ-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the γ-Tubulin complex. GCP2 (γ-Tubulin complex component 2), also known as TUBGCP2 or SPBC97 (spindle pole body protein Spc97 homolog) is a ubiquitously expressed 902 amino acid protein that localizes to the centrosome and is involved in microtubule nucleation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: component of cytoplasmic gamma-tubulin complex; Gamma-ring complex protein 103 kDa; Gamma-tubulin complex component 2; GCP-2; h103p; hGrip103; hSpc97; Spindle pole body protein Spc97 homolog; tubulin gamma complex associated protein 2; unnamed protein product

          View more View less

          Gene Aliases: ALP4; GCP-2; GCP2; Grip103; h103p; hGCP2; hSpc97; PAMDDFS; SPBC97; SPC97; Spc97p; TUBGCP2

          View more View less

          UniProt ID: (Human) Q9BSJ2

          View more View less

          Entrez Gene ID: (Human) 10844

          View more View less

          Function(s)
          protein binding gamma-tubulin binding microtubule minus-end binding
          Process(es)
          microtubule cytoskeleton organization mitotic cell cycle neuron migration microtubule nucleation brain development cytoplasmic microtubule organization spindle assembly meiotic cell cycle protein-containing complex assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline