Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TYK2 Antibodies

          Abnova

          TYK2 Polyclonal Antibody, MaxPab™

          View all (20) TYK2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TYK2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TYK2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TYK2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TYK2 Antibody (H00007297-D01P) in WB

          Western Blot analysis of TYK2 expression in transfected 293T cell line by TYK2 MaxPab polyclonal antibody. Lane 1: TYK2 transfected lysate(133.70 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TYK2 Antibody in Western Blot (WB)

          Product Details

          H00007297-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TYK2 (NP_003322.2, 1 a.a. approximately 1187 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPLRHWGMAR GSKPVGDGAQ PMAAMGGLKV LLHWAGPGGG EPWVTFSESS LTAEEVCIHI AHKVGITPPC FNLFALFDAQ AQVWLPPNHI LEIPRDASLM LYFRIRFYFR NWHGMNPREP AVYRCGPPGT EASSDQTAQG MQLLDPASFE YLFEQGKHEF VNDVASLWEL STEEEIHHFK NESLGMAFLH LCHLALRHGI PLEEVAKKTS FKDCIPRSFR RHIRQHSALT RLRLRNVFRR FLRDFQPGRL SQQMVMVKYL ATLERLAPRF GTERVPVCHL RLLAQAEGEP CYIRDSGVAP TDPGPESAAG PPTHEVLVTG TGGIQWWPVE EEVNKEEGSS GSSGRNPQAS LFGKKAKAHK AVGQPADRPR EPLWAYFCDF RDITHVVLKE HCVSIHRQDN KCLELSLPSR AAALSFVSLV DGYFRLTADS SHYLCHEVAP PRLVMSIRDG IHGPLLEPFV QAKLRPEDGL YLIHWSTSHP YRLILTVAQR SQAPDGMQSL RLRKFPIEQQ DGAFVLEGWG RSFPSVRELG AALQGCLLRA GDDCFSLRRC CLPQPGETSN LIIMRGARAS PRTLNLSQLS FHRVDQKEIT QLSHLGQGTR TNVYEGRLRV EGSGDPEEGK MDDEDPLVPG RDRGQELRVV LKVLDPSHHD IALAFYETAS LMSQVSHTHL AFVHGVCVRG PENIMVTEYV EHGPLDVWLR RERGHVPMAW KMVVAQQLAS ALSYLENKNL VHGNVCGRNI LLARLGLAEG TSPFIKLSDP GVGLGALSRE ERVERIPWLA PECLPGGANS LSTAMDKWGF GATLLEICFD GEAPLQSRSP SEKEHFYQRQ HRLPEPSCPQ LATLTSQCLT YEPTQRPSFR TILRDLTRLQ PHNLADVLTV NPDSPASDPT VFHKRYLKKI RDLGEGHFGK VSLYCYDPTN DGTGEMVAVK ALKADCGPQH RSGWKQEIDI LRTLYHEHII KYKGCCEDQG EKSLQLVMEY VPLGSLRDYL PRHSIGLAQL LLFAQQICEG MAYLHSQHYI HRDLAARNVL LDNDRLVKIG DFGLAKAVPE GHEYYRVRED GDSPVFWYAP ECLKEYKFYY ASDVWSFGVT LYELLTHCDS SQSPPTKFLE LIGIAQGQMT VLRLTELLER GERLPRPDKC PCEVYHLMKN CWETEASFRP TFENLIPILK TVHEKYQGQA PSVFSVC

          Target Information

          Tyrosine Kinase 2 (TYK2) a member of the JAK kinase family, plays both structural and catalytic roles in type I interferon (IFN) signaling. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; mKIAA4116; Non-receptor tyrosine-protein kinase TYK2; tyrosine kinase TYK2; unnamed protein product

          View more View less

          Gene Aliases: IMD35; JTK1; TYK2

          View more View less

          UniProt ID: (Human) P29597, (Mouse) Q9R117

          View more View less

          Entrez Gene ID: (Human) 7297, (Mouse) 54721

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity growth hormone receptor binding protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups type 1 angiotensin receptor binding hormone receptor binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter protein phosphorylation immune response JAK-STAT cascade cell proliferation cytokine-mediated signaling pathway regulation of cell-cell adhesion cell differentiation positive regulation of interferon-gamma production positive regulation of interleukin-17 production positive regulation of natural killer cell proliferation intracellular signal transduction interleukin-12-mediated signaling pathway interleukin-23-mediated signaling pathway type III interferon-mediated signaling pathway positive regulation of T cell proliferation regulation of JAK-STAT cascade positive regulation of JAK-STAT cascade regulation of alpha-beta T cell activation positive regulation of NK T cell proliferation interferon-gamma-mediated signaling pathway type I interferon signaling pathway JAK-STAT cascade involved in growth hormone signaling pathway cellular response to virus interleukin-10-mediated signaling pathway positive regulation of protein localization to nucleus positive regulation of T-helper 17 type immune response transmembrane receptor protein tyrosine kinase signaling pathway phosphorylation cell migration peptidyl-tyrosine phosphorylation peptidyl-tyrosine autophosphorylation regulation of cell proliferation innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline