Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GNSCRQEDV IATANLSRRA IADMLRACKE AAYHPEVAPD VRLRALHYGR ECANGYLELL DHVLLTLQKP SPELKQQLTG HSKRVAGSVT ELIQAAEAMK GTEWVDPEDP TVIAENELLG AAAAIEAAAK KLEQLKPRAK PKEADESLNF EEQILEAAKS IAAATSALVK AASAAQRELV AQGKVGAIPA NALDDGQWSQ GLISAARMVA AATNNLCEAA NAAVQGHASQ EKLISSAKQV AASTAQLLVA CKVKADQDSE AMKRLQAAGN AVKRASDNLV KAAQKAAAFE EQENETVVVK EKMVGGIAQI IAAQEEMLRK ERELEEARKK LAQIRQQQYK FLPSELRDEH (2193-2541 aa encoded by BC042923)
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoskeletal protein; unnamed protein product
Gene Aliases: ILWEQ; talin-1; TLN
UniProt ID: (Human) Q9Y490, (Mouse) P26039
Entrez Gene ID: (Human) 7094, (Rat) 313494, (Mouse) 21894
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support