Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • UACA Antibodies

          Abnova

          UACA Polyclonal Antibody, MaxPab™

          View all (13) UACA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UACA Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          UACA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          UACA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UACA Antibody (H00055075-B03P) in WB

          Western Blot analysis of UACA expression in transfected 293T cell line by UACA MaxPab polyclonal antibody. Lane 1: UACA transfected lysate(162.50 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UACA Antibody in Western Blot (WB)
          UACA Polyclonal Antibody, MaxPab™

          Product Details

          H00055075-B03P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UACA (NP_060473.2, 1 a.a. approximately 1416 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MKSLKSRLRR QDVPGPASSG AAAASAHAAD WNKYDDRLMK AAERGDVEKV TSILAKKGVN PGKLDVEGRS VFHVVTSKGN LECLNAILIH GVDITTSDTA GRNALHLAAK YGHALCLQKL LQYNCPTEHA DLQGRTALHD AAMADCPSSI QLLCDHGASV NAKDVDGRTP LVLATQMSRP TICQLLIDRG ADVNSRDKQN RTALMLGCEY GCRDAVEVLI KNGADISLLD ALGHDSSYYA RIGDNLDILT LLKTASENTN KGRELWKKGP SLQQRNLTHM QDEVNVKSHQ REHQNIQDLE IENEDLKERL RKIQQEQRIL LDKVNGLQLQ LNEEVMVADD LESEREKLKS LLAAKEKQHE ESLRTIEALK NRFKYFESDH LGSGSHFSNR KEDMLLKQGQ MYMADSQCTS PGIPAHMQSR SMLRPLELSL PSQTSYSENE ILKKELEAMR TFCESAKQDR LKLQNELAHK VAECKALALE CERVKEDSDE QIKQLEDALK DVQKRMYESE GKVKQMQTHF LALKEHLTSE AASGNHRLTE ELKDQLKDLK VKYEGASAEV GKLRNQIKQN EMIVEEFKRD EGKLIEENKR LQKELSMCEM EREKKGRKVT EMEGQAKELS AKLALSIPAE KFENMKSSLS NEVNEKAKKL VEMEREHEKS LSEIRQLKRE LENVKAKLAQ HVKPEEHEQV KSRLEQKSGE LGKKITELTL KNQTLQKEIE KVYLDNKLLK EQAHNLTIEM KNHYVPLKVS EDMKKSHDAI IDDLNRKLLD VTQKYTEKKL EMEKLLLEND SLSKDVSRLE TVFVPPEKHE KEIIALKSNI VELKKQLSEL KKKCGEDQEK IHALTSENTN LKKMMSNQYV PVKTHEEVKM TLNDTLAKTN RELLDVKKKF EDINQEFVKI KDKNEILKRN LENTQNQIKA EYISLAEHEA KMSSLSQSMR KVQDSNAEIL ANYRKGQEEI VTLHAEIKAQ KKELDTIQEC IKVKYAPIVS FEECERKFKA TEKELKDQLS EQTQKYSVSE EEVKKNKQEN DKLKKEIFTL QKDLRDKTVL IEKSHEMERA LSRKTDELNK QLKDLSQKYT EVKNVKEKLV EENAKQTSEI LAVQNLLQKQ HVPLEQVEAL KKSLNGTIEN LKEELKSMQR CYEKEQQTVT KLHQLLENQK NSSVPLAEHL QIKEAFEKEV GIIKASLREK EEESQNKMEE VSKLQSEVQN TKQALKKLET REVVDLSKYK ATKSDLETQI SSLNEKLANL NRKYEEVCEE VLHAKKKEIS AKDEKELLHF SIEQEIKDQK ERCDKSLTTI TELQRRIQES AKQIEAKDNK ITELLNDVER LKQALNGLSQ LTYTSGNPTK RQSQLIDTLQ HQVKSLEQQL ADADRQHQEV IAIYRTHLLS AAQGHMDEDV QEALLQIIQM RQGLVC

          Target Information

          Regulates APAF1 expression and plays an important role in the regulation of stress-induced apoptosis. Promotes apoptosis by regulating three pathways, apoptosome up-regulation, LGALS3/galectin-3 down-regulation and NF-kappa-B inactivation. Regulates the redistribution of APAF1 into the nucleus after proapoptotic stress. Down-regulates the expression of LGALS3 by inhibiting NFKB1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: nuclear membrane binding protein; unnamed protein product; Uveal autoantigen with coiled-coil domains and ankyrin repeats

          View more View less

          Gene Aliases: KIAA1561; NUCLING; UACA

          View more View less

          UniProt ID: (Human) Q9BZF9

          View more View less

          Entrez Gene ID: (Human) 55075

          View more View less

          Function(s)
          actin binding protein binding
          Process(es)
          intrinsic apoptotic signaling pathway in response to DNA damage intrinsic apoptotic signaling pathway in response to oxidative stress negative regulation of inflammatory response negative regulation of NIK/NF-kappaB signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline