Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • UBXN6 Antibodies

          Proteintech

          UBXD1 Polyclonal Antibody

          4 References
          View all (4) UBXN6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UBXD1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (23)
          UBXD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UBXD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UBXD1 Antibody (14706-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using 14706-1-AP (UBXD1 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UBXD1 Antibody in Immunocytochemistry (ICC/IF)
          UBXD1 Antibody in Immunocytochemistry (ICC/IF)
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBXD1 Antibody in Western Blot (WB)
          UBXD1 Antibody in Western Blot (WB)
          UBXD1 Antibody in Western Blot (WB)
          UBXD1 Polyclonal Antibody

          Product Details

          14706-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UBXD1 Fusion Protein Ag6394 (49-388 aa encoded by BC008288)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.16 mg/mL

          Amount

          24 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TNVVSEPRE EGSAHLAVPG VYFTCPLTGA TLRKDQRDAC IKEAILLHFS TDPVAASIMK IYTFNKDQDR VKLGVDTIAK YLDNIHLHPE EEKYRKIKLQ NKVFQERINC LEGTHEFFEA IGFQKVLLPA QDQEDPEEFY VLSETTLAQP QSLERHKEQL LAAEPVRAKL DRQRRVFQPS PLASQFELPG DFFNLTAEEI KREQRLRSEA VERLSVLRTK AMREKEEQRG LRKYNYTLLR VRLPDGCLLQ GTFYARERLG AVYGFVREAL QSDWLPFELL ASGGQKLSED ENLALNECGL VPSALLTFSW DMAVLEDIKA AGAEPDSILK PELLSAIEKL L (49-388 aa encoded by BC008288)

          Target Information

          May negatively regulate the ATPase activity of VCP, an ATP-driven segregase that associates with different cofactors to control a wide variety of cellular processes (PubMed:26475856). As a cofactor of VCP, it may play a role in the transport of CAV1 to lysosomes for degradation (PubMed:21822278, PubMed:23335559). It may also play a role in endoplasmic reticulum-associated degradation (ERAD) of misfolded proteins (PubMed:19275885). Together with VCP and other cofactors, it may play a role in macroautophagy, regulating for instance the clearance of damaged lysosomes (PubMed:27753622). [UniProt]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CTB-50L17.16; UBX domain containing 1; UBX domain-containing 2; UBX domain-containing protein 1; UBX domain-containing protein 6

          View more View less

          Gene Aliases: 1200008L11Rik; 2210415J11Rik; AU040909; UBXD1; UBXDC2; UBXN6

          View more View less

          UniProt ID: (Human) Q9BZV1, (Mouse) Q99PL6

          View more View less

          Entrez Gene ID: (Human) 80700, (Mouse) 66530, (Rat) 363332

          View more View less

          Function(s)
          protein binding ATPase binding
          Process(es)
          response to stress macroautophagy endosome to lysosome transport via multivesicular body sorting pathway ERAD pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline