Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • UGT1A7 Antibodies

          Abnova

          UGT1A7 Polyclonal Antibody, MaxPab™

          View all (2) UGT1A7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UGT1A7 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          UGT1A7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          UGT1A7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UGT1A7 Antibody (H00054577-B01P) in WB

          Western Blot analysis of UGT1A7 expression in transfected 293T cell line by UGT1A7 MaxPab polyclonal antibody. Lane 1: UGT1A7 transfected lysate(58.3 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UGT1A7 Antibody in Western Blot (WB)
          UGT1A7 Polyclonal Antibody, MaxPab™

          Product Details

          H00054577-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UGT1A7 (AAI53173.1, 1 a.a. approximately 530 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MARAGWTGLL PLYVCLLLTC GFAKAGKLLV VPMDGSHWFT MQSVVEKLIL RGHEVVVVMP EVSWQLGRSL NCTVKTYSTS YTLEDQDREF MVFADARWTA PLRSAFSLLT SSSNGIFDLF FSNCRSLFND RKLVEYLKES CFDAVFLDPF DACGLIVAKY FSLPSVVFAR GIFCHYLEEG AQCPAPLSYV PRLLLGFSDA MTFKERVWNH IMHLEEHLFC PYFFKNVLEI ASEILQTPVT AYDLYSHTSI WLLRTDFVLE YPKPVMPNMI FIGGINCHQG KPVPMEFEAY INASGEHGIV VFSLGSMVSE IPEKKAMAIA DALGKIPQTV LWRYTGTRPS NLANNTILVK WLPQNDLLGH PMTRAFITHA GSHGVYESIC NGVPMVMMPL FGDQMDNAKR METKGAGVTL NVLEMTSEDL ENALKAVIND KSYKENIMRL SSLHKDRPVE PLDLAVFWVE FVMRHKGAPH LRPAAHDLTW YQYHSLDVIG FLLAVVLTVA FITFKCCAYG YRKCLGKKGR VKKAHKSKTH

          Target Information

          UGT1A7 is an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The enzyme encoded by this gene has moderate glucuronidase activity with phenols.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Bilirubin-specific UDPGT isozyme 1; UDP glucuronosyltransferase 1 family, polypeptide A7; UDP glycosyltransferase 1 family, polypeptide A7; UDP-glucuronosyltransferase 1 family polypeptide A7s; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-7; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1-G; UDP-glucuronosyltransferase 1A1; UDP-glucuronosyltransferase 1A7; UDPGT 1-7; UGT-1G; UGT1A7

          View more View less

          Gene Aliases: GNT1; hUG-BR1; UDPGT; UDPGT 1-7; UGT-1A; UGT-1G; UGT1; UGT1-01; UGT1-07; UGT1.1; UGT1.7; UGT1A; UGT1A1; UGT1A7; UGT1G

          View more View less

          UniProt ID: (Human) Q9HAW7

          View more View less

          Entrez Gene ID: (Human) 54577

          View more View less

          Function(s)
          retinoic acid binding enzyme inhibitor activity protein kinase C binding protein binding UDP-glycosyltransferase activity glucuronosyltransferase activity transferase activity transferase activity, transferring glycosyl groups enzyme binding protein homodimerization activity protein heterodimerization activity
          Process(es)
          liver development lipid metabolic process fatty acid metabolic process xenobiotic metabolic process estrogen metabolic process toxin catabolic process coumarin metabolic process flavonoid metabolic process xenobiotic catabolic process retinoic acid metabolic process negative regulation of fatty acid metabolic process flavone metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-78576c459-66pvs:80/100.66.76.150:80.
          git-commit: 65c7ea78bfc366f1196f6925435a6e3b75c9e6fa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline