Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody specifically recognizes the endogenous UGT8.
Immunogen sequence: FKTLASALH ERGHHTVFLL SEGRDIAPSN HYSLQRYPGI FNSTTSDAFL QSKMRNIFSG RLTAIELFDI LDHYTKNCDL MVGNHALIQG LKKEKFDLLL VDPNDMCGFV IAHLLGVKYA VFSTGLWYPA EVGAPAPLAY VPEFNSLLTD RMNLLQRMKN TGVYLISRLG VSFLVLPKYE RIMQKYNLLP EKSMYDLVHG SSLWMLCTDV ALEFPRPTLP NVVYVGGILT KPASPLPEDL QRWVNGANEH GFVLVSFGAG VKYLSEDIAN KLAGALGRLP QKVIWRFSGP KPKNLGNNTK LIEWLPQNDL LGHSKIKAFL SHGGLNSIFE TMYHGVPVVG IPLFGDHYDT MTRVQAKGMG ILLEWK (38-402 aa encoded by BC075069)
UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ceramide UDP-galactosyltransferase; cerebroside synthase; UDP galactosyltransferase 8; UDP galactosyltransferase 8A; UDP-galactose-ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase 8; UDP-galactose-ceramide galactosyltransferase 8A; UDP-glucuronosyltransferase 8; unnamed protein product; uridine diphosphate glycosyltransferase 8
Gene Aliases: CGalT; CGT; mCerGT; UGT4; Ugt8; Ugt8a
UniProt ID: (Human) Q16880, (Rat) Q09426, (Mouse) Q64676
Entrez Gene ID: (Human) 7368, (Rat) 50555, (Mouse) 22239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support