          • Primary Antibodies ›
          • PSMA Antibodies
          OriGene
          FOLH1 Monoclonal Antibody (UMAB25), UltraMAB™
          View all (50) PSMA antibodies
          Tech Support
          Cite FOLH1 Monoclonal Antibody (UMAB25), UltraMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          FOLH1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FOLH1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          FOLH1 Antibody (UM570025) in ICC/IF
          Immunofluorescent staining of PC-12 cells using FOLH1 mouse monoclonal antibody (UM500025, green). Actin filaments were labeled with TRITC-phalloidin (red), and nuclear with DAPI (blue). The three-color overlay image is located at the bottom-right corner. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FOLH1 Antibody in Immunocytochemistry (ICC/IF)
          FOLH1 Antibody in Western Blot (WB)
          FOLH1 Antibody in Western Blot (WB)
          FOLH1 Antibody in Western Blot (WB)
          FOLH1 Antibody in Western Blot (WB)
          FOLH1 Monoclonal Antibody (UMAB25), UltraMAB™
          Product Details
          UM570025
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          ChIP assay (ChIP)
          Assay-Dependent
          -
          Product Specifications
          Species Reactivity
          Dog, Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          UMAB25
          Immunogen
          Full length human recombinant protein of human FOLH1 produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5-1.0 mg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a type II transmembrane glycoprotein belonging to the M28 peptidase family. The protein acts as a glutamate carboxypeptidase on different alternative substrates, including the nutrient folate and the neuropeptide N-acetyl-l-aspartyl-l-glutamate and is expressed in a number of tissues such as prostate, central and peripheral nervous system and kidney. A mutation in this gene may be associated with impaired intestinal absorption of dietary folates, resulting in low blood folate levels and consequent hyperhomocysteinemia. Expression of this protein in the brain may be involved in a number of pathological conditions associated with glutamate excitotoxicity. In the prostate the protein is up-regulated in cancerous cells and is used as an effective diagnostic and prognostic indicator of prostate cancer. This gene likely arose from a duplication event of a nearby chromosomal region. Alternative splicing gives rise to multiple transcript variants encoding several different isoforms.
          Bioinformatics
          Protein Aliases: Cell growth-inhibiting gene 27 protein; FGCP; Folate hydrolase 1; Folylpoly-gamma-glutamate carboxypeptidase; GCPII; GIG27; glutamate carboxylase II; Glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; glutamate carboxypeptidase II; folate hydrolase; Membrane glutamate carboxypeptidase; mGCP; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; NAALADase I; prostate specific membrane antigen; Prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSM-E; PSM-F; Pteroylpoly-gamma-glutamate carboxypeptidase; unnamed protein product
          View more View less
          Gene Aliases: FGCP; FOLH; FOLH1; GCP2; GCPII; GIG27; mGCP; Mopsm; Naalad; NAALAD1; PSM; PSMA
          View more View less
          UniProt ID: (Human) Q04609, (Mouse) O35409, (Rat) P70627
          View more View less
          Entrez Gene ID: (Human) 2346, (Dog) 476775, (Mouse) 53320, (Rat) 85309
          View more View less
          Function(s)
          catalytic activity carboxypeptidase activity metallocarboxypeptidase activity protein binding peptidase activity metallopeptidase activity hydrolase activity dipeptidase activity metal ion binding Ac-Asp-Glu binding tetrahydrofolyl-poly(glutamate) polymer binding metalloprotease
          Process(es)
          proteolysis glutamate biosynthetic process folic acid-containing compound metabolic process intestinal folate absorption positive regulation of glutamate receptor signaling pathway metabolic process positive regulation of apoptotic process
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline