Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • UNC45A Antibodies

          Abnova

          UNC45A Polyclonal Antibody, MaxPab™

          View all (3) UNC45A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UNC45A Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          UNC45A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          UNC45A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UNC45A Antibody (H00055898-B01P) in WB

          Western Blot analysis of UNC45A expression in transfected 293T cell line by UNC45A MaxPab polyclonal antibody. Lane 1: UNC45A transfected lysate(103.84 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UNC45A Antibody in Western Blot (WB)
          UNC45A Polyclonal Antibody, MaxPab™

          Product Details

          H00055898-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UNC45A (NP_061141, 1 a.a. approximately 944 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MTVSGPGTPE PRPATPGASS VEQLRKEGNE LFKCGDYGGA LAAYTQALGL DATPQDQAVL HRNRAACHLK LEDYDKAETE ASKAIEKDGG DVKALYRRSQ ALEKLGRLDQ AVLDLQRCVS LEPKNKVFQE ALRNIGGQIQ EKVRYMSSTD AKVEQMFQIL LDPEEKGTEK KQKASQNLVV LAREDAGAEK IFRSNGVQLL QRLLDMGETD LMLAALRTLV GICSEHQSRT VATLSILGTR RVVSILGVES QAVSLAACHL LQVMFDALKE GVKKGFRGKE GAIIVDPARE LKVLISNLLD LLTEVGVSGQ GRDNALTLLI KAVPRKSLKD PNNSLTLWVI DQGLKKILEV GGSLQDPPGE LAVTANSRMS ASILLSKLFD DLKCDAEREN FHRLCENYIK SWFEGQGLAG KLRAIQTVSC LLQGPCDAGN RALELSGVME SVIALCASEQ EEEQLVAVEA LIHAAGKAKR ASFITANGVS LLKDLYKCSE KDSIRIRALV GLCKLGSAGG TDFSMKQFAE GSTLKLAKQC RKWLCNDQID AGTRRWAVEG LAYLTFDADV KEEFVEDAAA LKALFQLSRL EERSVLFAVA SALVNCTNSY DYEEPDPKMV ELAKYAKQHV PEQHPKDKPS FVRARVKKLL AAGVVSAMVC MVKTESPVLT SSCRELLSRV FLALVEEVED RGTVVAQGGG RALIPLALEG TDVGQTKAAQ ALAKLTITSN PEMTFPGERI YEVVRPLVSL LHLNCSGLQN FEALMALTNL AGISERLRQK ILKEKAVPMI EGYMFEEHEM IRRAATECMC NLAMSKEVQD LFEAQGNDRL KLLVLYSGED DELLQRAAAG GLAMLTSMRP TLCSRIPQVT THWLEILQAL LLSSNQELQH RGAVVVLNMV EASREIASTL MESEMMEILS VLAKGDHSPV TRAAAACLDK AVEYGLIQPN QDGE

          Target Information

          UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GCUNC-45; general cell UNC45; Protein unc-45 homolog A; SMAP-1; smooth muscle cell associated protein-1; Smooth muscle cell-associated protein 1; unc-45 homolog A; Unc-45A; unnamed protein product

          View more View less

          Gene Aliases: GC-UNC45; GCUNC-45; GCUNC45; IRO039700; OOHE; SMAP-1; SMAP1; UNC-45A; UNC45A

          View more View less

          UniProt ID: (Human) Q9H3U1

          View more View less

          Entrez Gene ID: (Human) 55898

          View more View less

          Function(s)
          protein binding cadherin binding Hsp90 protein binding
          Process(es)
          protein folding muscle organ development cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline