Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • VAMP1/VAMP2 Antibodies

          Osenses

          VAMP 2,1 Polyclonal Antibody

          2 References
          View all (4) VAMP1/VAMP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VAMP 2,1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          VAMP 2,1 Polyclonal Antibody

          Product Details

          OSS00045G-500UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          10-50 µg/mL
          -

          Immunohistochemistry (IHC)

          10-50 µg/mL
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from aa region 10-60 of human VAMP2 conjugated to blue carrier protein was used as the antigen. The VAMP2 peptide shares 93% shared identity with VAMP1
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Amount

          500 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in many other species including rat, mouse and xenopus.

          Specificity of this antibody: VAMP2 and VAMP1.

          Target Information

          Vesicle-associated membrane protein 2 is essential for cAMP-regulated exocytosis. VAMP (also know as synaptobrevin) is part of the synaptic vesicle docking and fusion complex and plays a central role in neuroexocytosis. Two VAMP (vesicle-associated membrane protein) isoforms are expressed in the nervous system and are differently distributed among the specialized parts of the tissue. VAMP-1 and -2 are present in all tissues tested, including kidney, adrenal gland, liver, pancreas, thyroid, heart, and smooth muscle. The two isoforms are differentially expressed in various tissues and their level may depend on differentiation. VAMP-1 is restricted to exocrine pancreas and to kidney tubular cells, whereas VAMP-2 is the predominant isoform present in Langerhans islets and in glomerular cells. Both isoforms show a patchy vesicular intracellular distribution in confocal microscopy. Evidence for the importance of neuronal VAMPproteins in the physiology of all cells is indicated.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: spastic ataxia 1 (autosomal dominant); synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; Synaptobrevin-1; Synaptobrevin-2; unnamed protein product; VAMP-1; VAMP-2; VAMP1; vesicle targeting/fusion protein; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); Vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); Vesicle-associated membrane protein 2

          View more View less

          Gene Aliases: CMS25; lew; NEDHAHM; RATVAMPB; RATVAMPIR; SAX1; SPAX1; SYB; Syb-1; Syb-2; SYB1; SYB2; sybII; VAMP-1; VAMP-2; VAMP1; VAMP2

          View more View less

          UniProt ID: (Human) P23763, (Human) P63027, (Mouse) Q62442, (Rat) Q63666, (Mouse) P63044, (Rat) P63045

          View more View less

          Entrez Gene ID: (Human) 6843, (Human) 6844, (Mouse) 22317, (Rat) 25624, (Mouse) 22318, (Rat) 24803

          View more View less

          Function(s)
          SNAP receptor activity protein binding syntaxin binding SNARE binding calmodulin binding phospholipid binding syntaxin-1 binding calcium-dependent protein binding protein C-terminus binding myosin binding protein complex binding identical protein binding ion channel binding
          Process(es)
          vesicle fusion vesicle-mediated transport SNARE complex assembly exocytosis response to glucose protein transport synaptic vesicle exocytosis calcium ion regulated exocytosis regulation of exocytosis cellular response to insulin stimulus Golgi to plasma membrane protein transport eosinophil degranulation neutrophil degranulation natural killer cell degranulation synaptic vesicle endocytosis long-term synaptic potentiation regulation of vesicle-mediated transport membrane fusion protein-containing complex assembly mucus secretion positive regulation of intracellular protein transport regulation of delayed rectifier potassium channel activity regulation of histamine secretion by mast cell protein complex assembly regulation of calcium ion-dependent exocytosis neurotransmitter secretion intracellular protein transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline