Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • VAPA Antibodies

          Proteintech

          VAPA Polyclonal Antibody, CoraLite® Plus 488

          View all (21) VAPA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VAPA Polyclonal Antibody, CoraLite® Plus 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          VAPA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VAPA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VAPA Antibody (CL488-15275) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 VAPA antibody (CL488-15275) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VAPA Antibody in Immunocytochemistry (ICC/IF)
          VAPA Antibody in Flow Cytometry (Flow)

          Product Details

          CL488-15275

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          Flow Cytometry (Flow)

          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VAPA fusion protein Ag7393

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 0.5% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MASASGAMA KHEQILVLDP PTDLKFKGPF TDVVTTNLKL RNPSDRKVCF KVKTTAPRRY CVRPNSGIID PGSTVTVSVM LQPFDYDPNE KSKHKFMVQT IFAPPNTSDM EAVWKEAKPD ELMDSKLRCV FEMPNENDKL NDMEPSKAVP LNASKQDGPM PKPHSVSLND TETRKLMEEC KRLQGEMMKL SEENRHLRDE GLRLRKVAHS DKPGSTSTAS FR

          Target Information

          The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 33 kDa VAMP-associated protein; 33kDa (Vesicle Associated Membrane Protein)-associated protein; epididymis secretory sperm binding protein; synaptobrevin-associated protein of 33kDa; unnamed protein product; vamp; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP-A; VAMP-associated protein A; VAP-33; VAP-A; vesicle-associated membrane protein, associated protein a; Vesicle-associated membrane protein-associated protein A

          View more View less

          Gene Aliases: 33kDa; hVAP-33; VAMP-A; VAP-33; VAP-A; VAP33; VAPA

          View more View less

          UniProt ID: (Human) Q9P0L0, (Rat) Q9Z270, (Mouse) Q9WV55

          View more View less

          Entrez Gene ID: (Human) 9218, (Rat) 58857, (Mouse) 30960

          View more View less

          Function(s)
          protein binding microtubule binding protein domain specific binding FFAT motif binding protein homodimerization activity protein anchor cadherin binding protein heterodimerization activity signal transducer activity membrane traffic protein membrane trafficking regulatory protein
          Process(es)
          sphingomyelin biosynthetic process ER to Golgi vesicle-mediated transport endoplasmic reticulum organization phospholipid transport sterol transport viral release from host cell cholesterol transport neuron projection development protein folding in endoplasmic reticulum ceramide transport positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation by host of viral genome replication positive regulation by host of viral genome replication membrane fusion endoplasmic reticulum-plasma membrane tethering protein localization to endoplasmic reticulum COPII-coated vesicle budding signal transduction cell death positive regulation by host of viral release from host cell
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline