Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VEGFC Antibodies

          Proteintech

          VEGFC Polyclonal Antibody

          33 References
          View all (36) VEGFC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite VEGFC Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          VEGFC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VEGFC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VEGFC Antibody (22601-1-AP) in ICC/IF

          Immunofluorescent analysis of RAW 264.7 cells using 22601-1-AP (VEGFC antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VEGFC Antibody in Immunocytochemistry (ICC/IF)
          VEGFC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VEGFC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VEGFC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VEGFC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VEGFC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VEGFC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VEGFC Antibody in Western Blot (WB)
          VEGFC Antibody in Western Blot (WB)
          VEGFC Antibody in Western Blot (WB)
          VEGFC Polyclonal Antibody

          Product Details

          22601-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 18 publications 18 publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VEGFC Fusion Protein Ag18182 (179-228 aa encoded by BC035212)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Amount

          49.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: SYLSKTLFE ITVPLSQGPK PVTISFANHT SCRCMSKLDV YRQVHSIIRR S (179-228 aa encoded by BC035212)

          Target Information

          The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Flt4 ligand; FLT4 ligand DHM; Flt4-L; unnamed protein product; Vascular endothelial growth factor C; vascular endothelial growth factor C isoform 129; vascular endothelial growth factor C isoform 184; Vascular endothelial growth factor-related protein; VEGF-C; VEGF3; VEGFC; VRP

          View more View less

          Gene Aliases: AW228853; Flt4-L; LMPH1D; LMPHM4; VEGF-C; VEGFC; VRP

          View more View less

          UniProt ID: (Human) P49767, (Mouse) P97953

          View more View less

          Entrez Gene ID: (Human) 7424, (Mouse) 22341

          View more View less

          Function(s)
          protein binding growth factor activity chemoattractant activity vascular endothelial growth factor receptor 3 binding receptor agonist activity growth factor
          Process(es)
          angiogenesis response to hypoxia sprouting angiogenesis positive regulation of neuroblast proliferation substrate-dependent cell migration signal transduction cell proliferation positive regulation of cell proliferation response to xenobiotic stimulus organ morphogenesis glial cell proliferation morphogenesis of embryonic epithelium cell differentiation positive regulation of cell migration regulation of vascular endothelial growth factor receptor signaling pathway vascular endothelial growth factor signaling pathway positive regulation of blood vessel endothelial cell migration negative regulation of osteoblast differentiation positive regulation of angiogenesis negative regulation of blood pressure vascular endothelial growth factor receptor signaling pathway positive regulation of epithelial cell proliferation positive regulation of protein secretion positive chemotaxis induction of positive chemotaxis positive regulation of cell division positive regulation of glial cell proliferation positive regulation of mast cell chemotaxis positive regulation of lymphangiogenesis positive regulation of mesenchymal stem cell proliferation cellular response to leukemia inhibitory factor positive regulation of cell-matrix adhesion multicellular organism development negative regulation of cell proliferation positive regulation of protein autophosphorylation positive regulation of peptidyl-tyrosine phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-kvkbn:80/100.66.78.154:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline