Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FLK1 (VEGF Receptor 2) Antibodies

          AbboMax

          VEGFR-2 (CD309) Polyclonal Antibody

          View all (100) FLK1 (VEGF Receptor 2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VEGFR-2 (CD309) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          VEGFR-2 (CD309) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          VEGFR-2 (CD309) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VEGFR-2 (CD309) Antibody (602-090) in WB

          Western Blot: The whole lysate derived from Hela (20 µg/lane) immunoblotted by Rabbit anti – p53 (Cat# ) at 1:500. Observed a major immunoreactive band at molecular weight kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VEGFR-2 (CD309) Antibody in Western Blot (WB)
          VEGFR-2 (CD309) Polyclonal Antibody

          Product Details

          602-090

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2-10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to the epitope -KDYVG- without a Tyr951 phosphorylation site of human VEGFR-2 protein
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with proprietary stabilizer

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive control: bEnd.3 cells

          Cellular location: Membrane.

          Target Information

          KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD309; CD309 antigen; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); KDR; kinase insert domain protein receptor; Kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); Kinase NYK; Protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; soluble VEGFR2; tyrosine kinase growth factor receptor; unnamed protein product; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR-2; VEGFR-2; tyrosine kinase

          View more View less

          Gene Aliases: 6130401C07; CD309; Flk-1; FLK1; KDR; Krd-1; Ly73; sVEGFR-2; VEGFR; VEGFR-2; VEGFR2

          View more View less

          UniProt ID: (Human) P35968, (Rat) O08775, (Mouse) P35918

          View more View less

          Entrez Gene ID: (Human) 3791, (Rat) 25589, (Mouse) 16542

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity vascular endothelial growth factor-activated receptor activity integrin binding protein binding ATP binding coreceptor activity kinase activity transferase activity growth factor binding vascular endothelial growth factor binding identical protein binding cadherin binding Hsp90 protein binding transferase activity, transferring phosphorus-containing groups transmembrane signal receptor
          Process(es)
          angiogenesis ovarian follicle development patterning of blood vessels vasculogenesis positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation vasculature development lymph vessel development positive regulation of mesenchymal cell proliferation epithelial cell maturation endocardium development endothelium development transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cell proliferation regulation of cell shape mesenchymal cell proliferation positive regulation of endothelial cell migration negative regulation of gene expression positive regulation of macroautophagy cell migration peptidyl-tyrosine phosphorylation hemopoiesis cell differentiation lung development positive regulation of cell migration positive regulation of BMP signaling pathway embryonic hemopoiesis cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor receptor-2 signaling pathway vascular endothelial growth factor signaling pathway negative regulation of apoptotic process surfactant homeostasis regulation of MAPK cascade positive regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of blood vessel endothelial cell migration cell fate commitment endothelial cell differentiation positive regulation of angiogenesis protein autophosphorylation vascular endothelial growth factor receptor signaling pathway lung alveolus development post-embryonic camera-type eye morphogenesis branching morphogenesis of an epithelial tube epithelial cell proliferation positive regulation of epithelial cell proliferation positive regulation of positive chemotaxis positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling positive regulation of mitochondrial depolarization calcium ion homeostasis epithelial tube morphogenesis blood vessel endothelial cell differentiation vascular wound healing positive regulation of ERK1 and ERK2 cascade semaphorin-plexin signaling pathway stem cell proliferation positive regulation of cell migration involved in sprouting angiogenesis positive regulation of mitochondrial fission positive regulation of nitric oxide-cGMP mediated signal transduction regulation of hematopoietic progenitor cell differentiation regulation of bone development cellular response to hydrogen sulfide negative regulation of endothelial cell apoptotic process positive regulation of stem cell proliferation positive regulation of endothelial cell chemotaxis positive regulation of vasculogenesis response to hypoxia cell migration involved in sprouting angiogenesis endochondral bone growth positive regulation of cytosolic calcium ion concentration aging male gonad development positive regulation of phosphatidylinositol 3-kinase signaling hyaluronan metabolic process positive regulation of TOR signaling calcium-mediated signaling using intracellular calcium source peptidyl-tyrosine autophosphorylation response to drug regulation of endothelial cell differentiation positive regulation of vasodilation positive regulation of long-term neuronal synaptic plasticity cell maturation neuron projection morphogenesis positive regulation of neurogenesis positive regulation of calcium-mediated signaling positive regulation of nitric-oxide synthase biosynthetic process response to oxygen levels regulation of endothelial cell proliferation protein phosphorylation multicellular organism development phosphorylation positive regulation of vascular endothelial growth factor receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline