PA1-32574 detects VIPR1 from human samples.
PA1-32574 has been successfully used in immunohistochemistry (paraffin tissue) applications.
The PA1-32574 immunogen is: Synthetic peptide, KLH conjugated.
The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.