Search
Search
Osenses
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: VN1R3. See also the Applications.
Putative pheromone receptor. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC129709; MGC129710; similar to V1RL1; V1R-like 3; V1r-like receptor 3; V1RL3 pseudogene; vomeronasal 1 receptor 3 pseudogene; vomeronasal 1 receptor 4 pseudogene
Gene Aliases: FKSG46; V1RL3; V1RL3p
UniProt ID: (Human) Q9BXE9
Entrez Gene ID: (Human) 317702
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support