Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • VPS39 Antibodies

          Abnova

          VPS39 Polyclonal Antibody, MaxPab™

          View all (7) VPS39 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VPS39 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          VPS39 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          VPS39 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VPS39 Antibody (H00023339-B01P) in WB

          Western Blot analysis of VPS39 expression in transfected 293T cell line by VPS39 MaxPab polyclonal antibody. Lane1:VPS39 transfected lysate(96.25 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VPS39 Antibody in Western Blot (WB)
          VPS39 Polyclonal Antibody, MaxPab™

          Product Details

          H00023339-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VPS39 (NP_056104.2, 1 a.a. approximately 875 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MHDAFEPVPI LEKLPLQIDC LAAWEEWLLV GTKQGHLLLY RIRKDVGCNR FEVTLEKSNK NFSKKIQQIH VVSQFKILVS LLENNIYVHD LLTFQQITTV SKAKGASLFT CDLQHTETGE EVLRMCVAVK KKLQLYFWKD REFHELQGDF SVPDVPKSMA WCENSICVGF KRDYYLIRVD GKGSIKELFP TGKQLEPLVA PLADGKVAVG QDDLTVVLNE EGICTQKCAL NWTDIPVAME HQPPYIIAVL PRYVEIRTFE PRLLVQSIEL QRPRFITSGG SNIIYVASNH FVWRLIPVPM ATQIQQLLQD KQFELALQLA EMKDDSDSEK QQQIHHIKNL YAFNLFCQKR FDESMQVFAK LGTDPTHVMG LYPDLLPTDY RKQLQYPNPL PVLSGAELEK AHLALIDYLT QKRSQLVKKL NDSDHQSSTS PLMEGTPTIK SKKKLLQIID TTLLKCYLHT NVALVAPLLR LENNHCHIEE SEHVLKKAHK YSELIILYEK KGLHEKALQV LVDQSKKANS PLKGHERTVQ YLQHLGTENL HLIFSYSVWV LRDFPEDGLK IFTEDLPEVE SLPRDRVLGF LIENFKGLAI PYLEHIIHVW EETGSRFHNC LIQLYCEKVQ GLMKEYLLSF PAGKTPVPAG EEEGELGEYR QKLLMFLEIS SYYDPGRLIC DFPFDGLLEE RALLLGRMGK HEQALFIYVH ILKDTRMAEE YCHKHYDRNK DGNKDVYLSL LRMYLSPPSI HCLGPIKLEL LEPKANLQAA LQVLELHHSK LDTTKALNLL PANTQINDIR IFLEKVLEEN AQKKRFNQVL KNLLHAEFLR VQEERILHQQ VKCIITEEKV CMVCKKKIGN SAFARYPNGV VVHYFCSKEV NPADT

          Target Information

          VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hVam6p; OTTMUSP00000023341; OTTMUSP00000023350; PI3K type 3; TRAP1-like protein; unnamed protein product; vacuolar protein sorting 39 homolog; Vam6/Vps39-like protein; VPS39, HOPS complex subunit

          View more View less

          Gene Aliases: hVam6p; KIAA0770; TLP; VAM6; VPS39

          View more View less

          UniProt ID: (Human) Q96JC1

          View more View less

          Entrez Gene ID: (Human) 23339

          View more View less

          Function(s)
          protein binding
          Process(es)
          intracellular protein transport autophagy endosome to lysosome transport protein transport vesicle-mediated transport endocytic recycling endosomal vesicle fusion regulation of SNARE complex assembly autophagosome-lysosome fusion late endosome to lysosome transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline