Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • VPS54 Antibodies

          Proteintech

          VPS54 Polyclonal Antibody

          2 References
          View all (7) VPS54 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VPS54 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          VPS54 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          VPS54 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VPS54 Antibody (13327-1-AP) in WB

          Mouse brain tissue were subjected to SDS PAGE followed by western blot with 13327-1-AP (VPS54 antibody) at dilution of 1:400 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VPS54 Antibody in Western Blot (WB)
          VPS54 Antibody in Western Blot (WB)
          VPS54 Antibody in Western Blot (WB)
          VPS54 Antibody in Western Blot (WB)
          VPS54 Antibody in Immunoprecipitation (IP)
          VPS54 Polyclonal Antibody

          Product Details

          13327-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VPS54 Fusion Protein Ag4123 (757-977 aa encoded by BC030275)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: CVDNIPSVT TDMLTRLSDL LKYFNSRSCQ LVLGAGALQV VGLKTITTKN LALSSRCLQL IVHYIPVIRA HFEARLPPKQ YSMLRHFDHI TKDYHDHIAE ISAKLVAIMD SLFDKLLSKY EVKAPVPSAC FRNICKQMTK MHEAIFDLLP EEQTQMLFLR INASYKLHLK KQLSHLNVIN DGGPQNGLVT ADVAFYTGNL QALKGLKDLD LNMAEIWEQK R (757-977 aa encoded by BC030275)

          Target Information

          This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Hepatocellular carcinoma protein 8; hVps54L; protein phosphatase 1, regulatory subunit 164; Tumor antigen HOM-HCC-8; Tumor antigen SLP-8p; Tumor antigen SLP-8p homolog; unnamed protein product; vacuolar protein sorting 54 homolog; Vacuolar protein sorting-associated protein 54; VPS54, GARP complex subunit; Vps54-like protein

          View more View less

          Gene Aliases: HCC8; hVps54L; mSLP8; PPP1R164; SLP-8p; VPS54; VPS54L; Vsp54; WR

          View more View less

          UniProt ID: (Human) Q9P1Q0, (Rat) Q9JMK8, (Mouse) Q5SPW0

          View more View less

          Entrez Gene ID: (Human) 51542, (Rat) 286932, (Mouse) 245944

          View more View less

          Function(s)
          protein binding syntaxin binding membrane traffic protein
          Process(es)
          in utero embryonic development apoptotic DNA fragmentation protein targeting to lysosome protein targeting to vacuole cellular calcium ion homeostasis Golgi to vacuole transport striated muscle contraction mitochondrion organization vacuole organization lysosomal transport neuromuscular synaptic transmission skeletal muscle tissue development protein localization gene expression ubiquitin homeostasis microglia differentiation protein transport respiratory electron transport chain sphingolipid catabolic process protein localization to cell surface limb morphogenesis post-embryonic forelimb morphogenesis synaptic transmission, glutamatergic regulation of growth retrograde transport, endosome to Golgi protein targeting to ER response to antibiotic spermatid differentiation skeletal muscle tissue growth astrocyte differentiation neuron projection morphogenesis homeostasis of number of cells homeostasis of number of cells within a tissue musculoskeletal movement neuron apoptotic process response to calcium ion synaptic transmission, GABAergic L-glutamate import neurofilament cytoskeleton organization limb development motor behavior thrombin receptor signaling pathway cellular response to progesterone stimulus vesicle-mediated cholesterol transport motor neuron apoptotic process neural tissue regeneration neuroinflammatory response negative regulation of motor neuron apoptotic process growth transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline