PA1-74053 detects von Willebrand Factor from human samples.
PA1-74053 has been successfully used in ELISA and immunoelectrophoresis applications.
The PA1-74053 immunogen is von Willebrand factor from human plasma.
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.