Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Proteins & Peptides ›
          • WNT3A Proteins

          Gibco

          Wnt Surrogate-Fc Fusion Recombinant Protein

          1 Reference
          View all (2) WNT3A proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Wnt Surrogate-Fc Fusion Recombinant Protein

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (7)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Wnt Surrogate-Fc Fusion Protein (PHG0402) in Bioactivity

          The Wnt surrogate transcriptome is highly similar to Wnt3a-regulated transcriptomes. Untreated iPSCs were cultured in DMEM/F-12 in the absence of Wnt agonist; StemFlex Medium is the PSC culture medium. All conditions were assayed 3 times as indicated by the labels on the left of the heat map. The heat map contains 4,744 genes with log2(rpm+1) and > 2.5-fold change in gene expression threshold; rpm = reads p... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)
          Wnt Surrogate-Fc Fusion Protein in Bioactivity (Bioactivity)

          Product Details

          PHG0402

          Applications
          Tested Dilution
          Publications

          Bioactivity (Bioactivity)

          Assay-dependent
          View 1 publication 1 publication
          Product Specifications

          Species

          Human

          Published species

          Human

          Expression System

          HEK293 cells

          Class

          Recombinant

          Type

          Protein

          Purity

          > 95%

          Endotoxin concentration

          1 EU/mL

          Activity

          Greater than 25 fold induction in Topflash assay activity at a Wnt surrogate-Fc protein concentration between 0.03 - 5.0 nM (based on dimeric stoichiometry)

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          25mM tris, pH 8.2, with 500mM NaCl

          Contains

          no preservative

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Dry ice

          Product Specific Information

          Gibco Wnt Surrogate-Fc Fusion Protein activates similar Wnt signaling pathways as most commercially available recombinant Wnt3a proteins or Wnt-conditioned media, while requiring a fraction of the amount of protein or hassle. Gibco Wnt Surrogate-Fc Fusion Protein was engineered with improved solubility, overcoming a major obstacle of working with Wnt3a and reducing the concentrations required to activate Wnt signaling in your model system.

          Gibco Wnt Surrogate-Fc Fusion Protein is comprised of the LRP-binding domain of DKK linked to a selective, high affinity binder of Frizzled receptors.

          Gibco Wnt Surrogate-Fc Fusion Protein is provided at a concentration of 2 mg/mL which is equivalent to 17.1 µM based on dimeric stoichiometry.

          Storage: Store undiluted Gibco Wnt Surrogate-Fc Fusion Protein at -80° for up to 1 year. After thawing, apportion into working aliquots and store at -20°C to -80°C for up to 2 months.

          Target Information

          Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MGC119418; MGC119419; MGC119420; Protein Wnt-3a; unnamed protein product; wingless-type MMTV integration site family, member 3A; Wnt 3a; WNT3A protein

          View more View less

          Gene Aliases: WNT3A

          View more View less

          UniProt ID: (Human) P56704

          View more View less

          Entrez Gene ID: (Human) 89780

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0