Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAQTSPAEK GLNPGLMCQE SYACSGTDEA IFECDECCSL QCLRCEEELH RQERLRNHER IRLKPGHVPY CDLCKGLSGH LPGVRQRAIV RCQTCKINLC LECQKRTHSG GNKRRHPVTV YNVSNLQESL EAEEMDEETK RKKMTEKVVS FLLVDENEEI QVTNEEDFIR KLDCKPDQHL KVVSIFGNTG DGKSHTLNHT FFYGREVFKT SPTQESCTVG VWAAYDPVHK VAVIDTEGLL GATVNLSQRT RLLLKVLAIS DLVIYRTHAD RLHNDLFKFL GDASEAYLKH FTKELKATTA RCGLDVPLST LGPAVIIFHE TVHTQLLGSD HPSEVPEKLI QDRFRKLGRF PEAFSSIHYK GTRTYNPPTD FSGLRRALEQ LLENNTTRSP RHPGVIFKAL KALSDRFSGE IPDDQMAHSS FFPDEYFTCS SLCLSCGVGC KKSMNHGKEG VPHEAKSRCR YSHQYDNRVY TCKACYERGE EVSVVPKTSA STDSPWMGLA KYAWSGYVIE CPNCGVVYRS RQYWFGNQDP VDTVVRTEIV HVWPGTDGFL KDNNNAAQRL LDGMNFMAQS VSELSLGPTK AVTSWLTDQI APAYWRPNSQ ILSCNKCATS FKDNDTKHHC RACGEGFCDS CSSKTRPVPE RGWGPAPVRV CDNCYEARNV QLAVTEAQVD DEGGTLIARK VGEAVQNTLG AVVTAIDIPL GLVKDAARPA YWVPDHEILH CHNCRKEFSI KLSKHHCRAC GQGFCDECSH DRRAVPSRGW DHPVRVCFNC NKKPGDL
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DFCP1; Double FYVE-containing protein 1; phosphoinositide-binding protein SR3; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; unnamed protein product; Zinc finger FYVE domain-containing protein 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1
Gene Aliases: DFCP1; KIAA1589; PP10436; PPP1R172; SR3; TAFF1; ZFYVE1; ZNFN2A1
UniProt ID: (Human) Q9HBF4
Entrez Gene ID: (Human) 53349
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support