Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ZIC Pan Antibodies

          Invitrogen

          ZIC Pan Polyclonal Antibody

          View all (2) ZIC Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZIC Pan Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          ZIC Pan Polyclonal Antibody

          Product Details

          PA5-68610

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.125 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          synthetic peptide directed towards the N terminal region of human ZIC Pan
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 2% sucrose

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2688572

          Product Specific Information

          This target displays homology in the following species: Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 100%; Zebrafish: 79%

          Target Information

          This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq, Jul 2008].

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: heterotaxy 1; transcription factor; unnamed protein product; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3 (odd-paired homolog, Drosophila); Zic family member 5 (odd-paired homolog, Drosophila); ZIC5; zinc family member 4 protein HZIC4; Zinc finger protein 201; Zinc finger protein 203; Zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; Zinc finger protein of the cerebellum 3; Zinc finger protein of the cerebellum 4; Zinc finger protein of the cerebellum 5; Zinc finger protein ZIC 1; Zinc finger protein ZIC 2; Zinc finger protein ZIC 3; Zinc finger protein ZIC 4; Zinc finger protein ZIC 5

          View more View less

          Gene Aliases: BAIDCS; CRS6; HPE5; HTX; HTX1; VACTERLX; ZIC; ZIC1; ZIC2; ZIC3; ZIC4; ZIC5; ZNF201; ZNF203

          View more View less

          UniProt ID: (Human) Q15915, (Human) O95409, (Human) O60481, (Human) Q8N9L1, (Human) Q96T25

          View more View less

          Entrez Gene ID: (Human) 7545, (Human) 7546, (Human) 7547, (Human) 84107, (Human) 85416

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding metal ion binding sequence-specific double-stranded DNA binding chromatin DNA binding transcription coactivator activity sequence-specific DNA binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter central nervous system development nervous system development brain development visual perception cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of sequence-specific DNA binding transcription factor activity skeletal system development neural plate development heart looping atrial cardiac muscle tissue development smoothened signaling pathway determination of left/right symmetry gastrulation pattern specification process mesoderm development heart development embryonic pattern specification anterior/posterior pattern specification gene expression stem cell population maintenance hippocampus development olfactory bulb development neuron differentiation lung development germ-line stem cell population maintenance forebrain development limb morphogenesis central nervous system segmentation determination of pancreatic left/right asymmetry determination of left/right asymmetry in nervous system outer ear morphogenesis mRNA transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter axial mesoderm development paraxial mesoderm development stem cell differentiation face development left/right pattern formation left/right axis specification determination of digestive tract left/right asymmetry determination of liver left/right asymmetry primitive streak formation maintenance of cell number cranial skeletal system development adult walking behavior regulation of smoothened signaling pathway spinal cord development positive regulation of protein import into nucleus inner ear morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline