Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ZMPSTE24 Antibodies

          Invitrogen

          ZMPSTE24 Polyclonal Antibody

          View all (13) ZMPSTE24 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZMPSTE24 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          ZMPSTE24 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ZMPSTE24 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZMPSTE24 Antibody (PA5-116870) in IHC (P)

          Immunohistochemistry analysis of ZMPSTE24 in paraffin-embedded human colon. Samples were incubated with ZMPSTE24 Polyclonal antibody (Product # PA5-116870) using a dilution of 1:100 (40x lens). Perform microwave antigen retrieval with 10 mM PBS buffer pH 7.2 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZMPSTE24 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZMPSTE24 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZMPSTE24 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZMPSTE24 Antibody in Western Blot (WB)

          Product Details

          PA5-116870

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.81 mg/mL

          Amount

          181 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2901501

          Product Specific Information

          Positive Samples: SKOV3

          Target Information

          This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CAAX prenyl protease 1 homolog; FACE-1; Farnesylated proteins-converting enzyme 1; farnesylated-proteins converting enzyme 1; FLJ14968; Prenyl protein-specific endoprotease 1; unnamed protein product; zinc metallopeptidase STE24; zinc metallopeptidase STE24 homolog; zinc metallopeptidase, STE24 homolog; Zinc metalloproteinase Ste24 homolog; zinc metalloproteinase, STE24 homolog

          View more View less

          Gene Aliases: A530043O15Rik; D030046F19; FACE-1; FACE1; HGPS; MADB; PRO1; RSDM1; STE24; Ste24p; ZMPSTE24

          View more View less

          UniProt ID: (Human) O75844, (Mouse) Q80W54

          View more View less

          Entrez Gene ID: (Human) 10269, (Mouse) 230709, (Rat) 313564

          View more View less

          Function(s)
          double-stranded DNA binding endopeptidase activity metalloendopeptidase activity protein binding peptidase activity metalloexopeptidase activity metallopeptidase activity hydrolase activity metal ion binding molecular_function metalloprotease
          Process(es)
          liver development hair follicle development heart morphogenesis ventricular cardiac muscle tissue development cardiac ventricle development growth plate cartilage development DNA repair chromatin organization regulation of transcription, DNA-templated proteolysis lipid metabolic process inflammatory cell apoptotic process cellular response to DNA damage stimulus nucleus organization nuclear envelope organization regulation of mitotic cell cycle adult walking behavior regulation of heart contraction determination of adult lifespan regulation of cell shape epidermis development regulation of autophagy positive regulation of gene expression negative regulation of gene expression regulation of glucose metabolic process protein processing regulation of lipid metabolic process bone mineralization prenylated protein catabolic process regulation of bone mineralization regulation of TOR signaling regulation of hormone metabolic process multicellular organism growth regulation of multicellular organism growth regulation of gene expression, epigenetic maintenance of rDNA regulation of DNA damage response, signal transduction by p53 class mediator hypomethylation of CpG island regulation of fibroblast proliferation thymus development regulation of defense response to virus neuromuscular process chromosome organization protein maturation cardiac muscle cell development regulation of ventricular cardiac muscle cell membrane repolarization kidney morphogenesis cardiac conduction CAMKK-AMPK signaling cascade regulation of stress-activated protein kinase signaling cascade cellular response to gamma radiation CAAX-box protein processing response to DNA damage checkpoint signaling regulation of mitotic cell cycle DNA replication regulation of blood circulation negative regulation of production of miRNAs involved in gene silencing by miRNA calcium ion import into sarcoplasmic reticulum regulation of termination of RNA polymerase I transcription regulation of cellular senescence biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline