Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • ZMYND10 Antibodies

          Proteintech

          ZMYND10 Polyclonal Antibody

          3 References
          View all (5) ZMYND10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZMYND10 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          ZMYND10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ZMYND10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZMYND10 Antibody (14431-1-AP) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with (Product # 14431-1-AP) ZMYND10 antibody at dilution of 1:800 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZMYND10 Antibody in Western Blot (WB)
          ZMYND10 Antibody in Western Blot (WB)
          ZMYND10 Antibody in Immunoprecipitation (IP)
          ZMYND10 Polyclonal Antibody

          Product Details

          14431-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ZMYND10 Fusion Protein Ag5808 (1-440 aa encoded by BC033732)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          25.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGDLELLLP GEAEVLVRGL RSFPLREMGS EGWNQQHENL EKLNMQAILD ATVSQGEPIQ ELLVTHGKVP TLVEELIAVE MWKQKVFPVF CRVEDFKPQN TFPIYMVVHH EASIINLLET VFFHKEVCES AEDTVLDLVD YCHRKLTLLV AQSGCGGPPE GEGSQDSNPM QELQKQAELM EFEIALKALS VLRYITDCVD SLSLSTLSRM LSTHNLPCLL VELLEHSPWS RREGGKLQQF EGSRWHTVAP SEQQKLSKLD GQVWIALYNL LLSPEAQARY CLTSFAKGRL LKLRAFLTDT LLDQLPNLAH LQSFLAHLTL TETQPPKKDL VLEQIPEIWE RLERENRGKW QAIAKHQLQH VFSPSEQDLR LQARRWAETY RLDVLEAVAP ERPRCAYCSA EASKRCSRCQ NEWYCCRECQ VKHWEKHGKT CVLAAQGDRA K (1-440 aa encoded by BC033732)

          Target Information

          The ZMYND10 gene encodes a protein that contains a MYND-type zinc finger domain, playing a crucial role in the assembly and function of cilia. Specifically, ZMYND10 is essential for the proper axonemal assembly of both inner and outer dynein arms, which are critical components of motile cilia. Mutations in ZMYND10 are associated with primary ciliary dyskinesia (PCD), a genetic disorder characterized by defective ciliary motility leading to recurrent respiratory infections and male infertility. In model organisms like mice and Drosophila, ZMYND10 expression is restricted to tissues and cells with motile cilia, such as chordotonal sensory neurons and sperm cells. Additionally, ZMYND10 has been identified as an epigenetically regulated tumor suppressor in various cancers, with its downregulation affecting processes like cell proliferation and tumor progression. The gene's dual role in ciliary function and tumorigenesis underlines its importance in both normal cellular physiology and disease pathology.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ciliary dyskinesia, primary, 22; dynein axonemal assembly factor 7; Protein BLu; tumor suppressor BLU; unnamed protein product; Zinc finger MYND domain-containing protein 10; zinc finger, MYND domain-containing 10

          View more View less

          Gene Aliases: BLU; CILD22; DNAAF7; FLU; LUCA12.4; ZMYND10

          View more View less

          UniProt ID: (Human) O75800, (Mouse) Q99ML0, (Rat) Q6AXZ5

          View more View less

          Entrez Gene ID: (Human) 51364, (Mouse) 114602, (Rat) 363139

          View more View less

          Function(s)
          protein binding zinc ion binding protein binding involved in protein folding metal ion binding binding, bridging molecular_function
          Process(es)
          outer dynein arm assembly inner dynein arm assembly motile cilium assembly cilium movement protein folding protein localization to cilium positive regulation of motile cilium assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline