Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLENYRNLVF LGIAVSKPDL ITCLEQGKEP CNMKRHEMVA KPPVMCSHIA EDLCPERDIK YFFQKVILRR YDKCEHENLQ LRKGCKSVDE CKVCKGGYNG LNQCLITTQS KMYQCDKYVK VFYKFSNSDR HKIRHTEKKT CKCKECGKSF CMLSQLTRHK RIHIRENSHK CEECGKAFNQ SSALTRHKMT HTGEKPYKCE ECGKAFNRSS HLTQHKVIHT REKPYKCEEC GKAFNRSSHI TQHKRIHNRE KPFKYDECCK AFKWSSALTT LTQHKRIHTG EKPYKCEECG KAFNQSSALT RHKMIHTGEK PFQCEECGKA FNGSSHLTQH KIIHTKEKPY KCEECGKAFN RSSHLTKHKR IHTREKAYKC DEYCKAFNWS SALTTLTQHK IIHTGEKPYK CEECGKAFNR SSYLIRHKII HTGEKPYKCE ECGKAFNQSS HLTQHKIIHT GEKPYKCEEC GKAFNRSSHL SQHKIIHTGE KPYKCEECGK PFNRFSYLTV HKRIHAGENP NKYEECGKAC NHSSNLTKHN S
ZNF257 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMZF-4; Bone marrow zinc finger 4; unnamed protein product; Zinc finger protein 257
Gene Aliases: BMZF-4; BMZF4; ZNF257
UniProt ID: (Human) Q9Y2Q1
Entrez Gene ID: (Human) 113835
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support