Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IEFGRQQRPK VGCFSQMIFK KHKSLPLHKR NNTREKSYEC KEYKKGFRKY LHLTEHLRDH TGVIPYECN
Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 41%.
ZNF404 (Zinc Finger Protein 404) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support