Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSRPRFNPRG DFPLQRPRAP NPSGMRPPGP FMRPGSMGLP RFYPAGRARG IPHRFAGHES YQNMGPQRMN VQVTQHRTDP RLTKEKLDFH EAQQKKGKPH GSRWDDEPHI SASVAVKQSS VTQVTEQSPK VQSRYTKESA SSILASFGLS NEDLEELSRY PDEQLTPENM PLILRDIRMR KMGRRLPNLP SQSRNKETLG SEAVSSNVID YGHASKYGYT EDPLEVRIYD PEIPTDEVEN EFQSQQNISA SVPNPNVICN SMFPVEDVFR QMDFPGESSN NRSFFSVESG TKMSGLHISG; Positive Samples: Mouse brain; Cellular Location: Nucleus speckle
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. NP220 (Nuclear protein 220), also designated Zinc finger protein 638 (ZNF638) or Cutaneous T-cell lymphoma-associated antigen se33-1, is a 1978 amino acid protein that contains one matrin-type zinc finger and two RRM (RNA recognition motif) domains, suggesting a role in transcriptional regulation. NP220 binds to double-stranded DNA fragments by recognizing clusters of cytidines. NP220 interacts with FHL-2 and is also thought to be phosphorylated by ATM or ATR upon DNA damage. It exists as five isoforms as a result of alternative splicing events. Isoform five of NP220 is a tumor-associated antigen found in several cutaneous T-cell lymphoma (CTCL), and in particular in mycosis fungoides patients and in Sezary syndrome patients.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTCL tumor antigen se33-1; CTCL-associated antigen se33-1; cutaneous T-cell lymphoma-associated antigen se33-1; NP220 nuclear protein; nuclear protein 220; unnamed protein product; zinc finger matrin-like protein; zinc finger, matrin-like
Gene Aliases: mNP220; NP220; ZFML; Zfp638; Znf638
UniProt ID: (Human) Q14966, (Mouse) Q61464
Entrez Gene ID: (Human) 27332, (Mouse) 18139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support