Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LPVYDTLMF CASRNTDRIH IYTKDGKQMS CNFIPLDIKL DLWEDLPASF QLKQYRSLIL RFVREWSSLT AMKQRIIRKS GQLFCSPILA LEEITKQQTK QNCTKRYITK EDVAVASMDK VKNVGGHVRL ITKESRPRDP FTKKLLEDGA CVPFLNPYTV QADLTVKPST SKGYLQAVDN EGNPLCLRCQ QPTCQTKQAC KANSWDSRFC SLKCQEEFWI RSNNSYLRAK VFETEHGVCQ LCNVNAQELF LRLRDAPKSQ RKNLLYATWT SKLPLEQLNE MIRNPGEGHF WQVDHIKPVY GGGGQCSLDN LQTLCTVCHK ERTARQAKER SQVRRQSLAS KHGSDITRFL VKK (728-1079 aa encoded by B C064616)
DNA annealing helicase and endonuclease required to maintain genome stability at stalled or collapsed replication forks by facilitating fork restart and limiting inappropriate recombination that could occur during template switching events (PubMed:21078962, PubMed:22704558, PubMed:22705370, PubMed:22759634, PubMed:26884333). Recruited to the sites of stalled DNA replication by polyubiquitinated PCNA and acts as a structure-specific endonuclease that cleaves the replication fork D-loop intermediate, generating an accessible 3'-OH group in the template of the leading strand, which is amenable to extension by DNA polymerase (PubMed:22759634). In addition to endonuclease activity, also catalyzes the fork regression via annealing helicase activity in order to prevent disintegration of the replication fork and the formation of double-strand breaks (PubMed:22705370, PubMed:22704558). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: annealing helicase 2; unnamed protein product; zinc finger Ran-binding domain-containing protein 3; zinc finger, RAN-binding domain containing 3
Gene Aliases: 4933425L19Rik; AH2
UniProt ID: (Human) Q5FWF4
Entrez Gene ID: (Human) 84083
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support