Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody recognizes a 33 kDa protein in SDS page.
Immunogen sequence: MDNSGKEAE AMALLAEAER KVKNSQSFFS GLFGGSSKIE EACEIYARAA NMFKMAKNWS AAGNAFCQAA QLHLQLQSKH DAATCFVDAG NAFKKADPQE AINCLMRAIE IYTDMGRFTI AAKHHISIAE IYETELVDIE KAIAHYEQSA DYYKGEESNS SANKCLLKVA GYAALLEQYQ KAIDIYEQVG TNAMDSPLLK YSAKDYFFKA ALCHFCIDML NAKLAVQKYE ELFPAFSDSR ECKLMKKLLE AHEEQNVDSY TESVKEYDSI SRLDQWLTTM LLRIKKTIQG DEEDLR (1-295 aa encoded by B C007432)
SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs (SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha snap; alpha-SNAP; hydrocephaly with hop gait; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; SNAP-alpha; unnamed protein product
Gene Aliases: 1500039N14Rik; a-SNAP; alpha-SNAP; hyh; RA81; SNAP-alpha; SNAPA; SNARE
UniProt ID: (Human) P54920, (Mouse) Q9DB05, (Rat) P54921
Entrez Gene ID: (Human) 8775, (Mouse) 108124, (Rat) 140673
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support