Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • beta Amyloid Antibodies

          Invitrogen

          beta Amyloid Polyclonal Antibody (CT695)

          Advanced Verification
          80 Published Figures
          149 References
          View all (80) beta Amyloid antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite beta Amyloid Polyclonal Antibody (CT695)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (80)
          • Advanced Verification (2)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 85

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          beta Amyloid Antibody (51-2700) in IHC

          Rabbit anti-Beta-Amyloid Precursor Protein (CT695) stained rat corticospinal tract after 24 hrs Traumatic Axonal Injury. Image: Courtesy of Dr. James R. Stone, University of Virginia, Dept. of Neurosurgery. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          beta Amyloid Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          beta Amyloid Antibody in Immunocytochemistry (ICC/IF)
          beta Amyloid Antibody in Immunocytochemistry (ICC/IF)
          beta Amyloid Antibody in Immunocytochemistry (ICC/IF)
          beta Amyloid Antibody in Immunocytochemistry (ICC/IF)
          beta Amyloid Antibody in Immunocytochemistry (ICC/IF)
          beta Amyloid Antibody in Immunohistochemistry (Frozen) (IHC (F))
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry (IHC)
          beta Amyloid Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot (WB)
          beta Amyloid Antibody in Western Blot, Immunoprecipitation (WB, IP)
          beta Amyloid Antibody in Western Blot, Immunoprecipitation (WB, IP)
          beta Amyloid Antibody
          beta Amyloid Antibody
          beta Amyloid Polyclonal Antibody (CT695)

          Product Details

          51-2700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.2-2 µg/mL
          View 50 publications 50 publications

          Immunohistochemistry (IHC)

          0.1-1.0 µg/mL
          View 62 publications 62 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 8 publications 8 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          -
          View 7 publications 7 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rat

          Published species

          Hamster, Human, Mouse, Pig, Rat, Tag

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Clone

          CT695

          Immunogen

          A 22 amino acid synthetic peptide derived from the C-terminus of the human beta-amyloid precursor protein (beta-APP). This sequence is 100% conserved in human, monkey, mouse, rat, guinea pig, and chicken beta-APP proteins.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533902

          Product Specific Information

          This antibody can be used to specifically detect the beta-amyloid precursor protein. The antibody reacts with full-length (APP695, 751, 770) and N-terminal truncated forms of beta-APP. The antibody can also be used to detect the C-terminal membrane-anchored fragment of beta-APP that remains after alpha- or beta-secretase cleavage. This antibody does not detect the beta-APP product N-terminal to the gamma-secretase cleavage site.

          Reactivity with this antibody has been confirmed for human, mouse, pig, and rat. Based on sequence homology, reactivity with other species, including monkey, guinea pig, and chicken beta-APP, is highly likely. The CT695 antibody has demonstrated superiority in detecting axonal damage.

          This antibody has been tested in western blotting with FRTL-5 (rat thyroid) cells and rat brain lysates. For immunohistochemistry FFPE tissues will require epitope retrieval pretreatment.

          Target Information

          Amyloid beta peptide (Abeta/Beta-amyloid) is the major constituent of amyloid plaques in the brains of individuals afflicted with Alzheimer's disease. Abeta peptide is 40-43 amino acids long and generated from the beta-amyloid precursor protein (beta APP) in a two-step process. The first step involves cleavage of the extracellular, amino-terminal domain of beta APP. Protein cleavage is performed by an aspartyl protease, beta-secretase (BACE) which is synthesized as a propeptide and must be modified to the mature and active form by the prohormone convertase, furin. Beta APP cleavage by the mature form of BACE results in the cellular secretion of a segment of beta APP, and a membrane-bound remnant. The remnant protein is processed by another protease, gamma-secretase. Gamma-secretase cleaves an intra-membrane site in the carboxyl-terminal domain of beta APP, thus generating the amyloid beta peptide. Gamma-secretase is believed to be a multi-subunit complex containing presenilin-1 and 2 as central components. The transmembrane glycoprotein, nicastrin, is associated with presinilins and has been found to bind to the carboxyl-terminus of beta APP and helps to modulate the production of the amyloid beta peptide. Abeta is an extracellular filamentous protein component of amyloid cores, neuritic plaques and is also found as a deposit in neurofibrillary tangles. Alzheimer's disease, the most common cause of senile dementia, is characterized by abnormal filamentous protein deposits in the brain. Beta amyloid deposits are also detected in Lewy body dementia, Down's syndrome, amyloidosis (Dutch type), cerebroarterial amyloidosis (cerebral amyloid angiopathy) and in the Guam Parkinson-Dementia complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ABPP; AG; Alzheimer disease; Alzheimer disease amyloid A4 protein homolog; Alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; amyloid beta (A4) precursor protein (peptidase nexin-II, Alzheimer disease); amyloid beta A4 protein; amyloid beta precursor protein (ABPP); amyloid beta protein; amyloid beta-protein precursor; Amyloid precursor protein; amyloid protein; Amyloid β; Amyloid-beta (A4) precursor protein; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloidogenic glycoprotein; APP; APPI; appican; beta-amyloid A4; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; Cerebral vascular amyloid peptide; CTF gamma; CTF-alpha; CVAP; OTTHUMP00000096096; Pan-Abeta; peptidase nexin-II; PN-II; PreA4; PreA4 751; protease nexin II; Protease nexin-II; testicular tissue protein Li 2; unnamed protein product

          View more View less

          Gene Aliases: A4; AAA; ABETA; ABPP; AD1; Adap; Ag; alpha-sAPP; APP; APPI; betaApp; CTFgamma; CVAP; E030013M08Rik; PN-II; PN2; preA4

          View more View less

          UniProt ID: (Human) B2R5V1, (Pig) P79307, (Rat) P08592, (Mouse) P12023

          View more View less

          Entrez Gene ID: (Human) 351, (Pig) 397663, (Rat) 54226, (Mouse) 11820

          View more View less

          Function(s)
          G-protein coupled receptor binding DNA binding chromatin binding serine-type endopeptidase inhibitor activity receptor binding frizzled binding insulin receptor binding integrin binding protein binding heparin binding peptidase activator activity enzyme binding peptidase inhibitor activity receptor activator activity acetylcholine receptor activator activity acetylcholine receptor binding apolipoprotein binding chemoattractant activity identical protein binding protein homodimerization activity heparan sulfate proteoglycan binding metal ion binding ephrin receptor binding transition metal ion binding protein heterodimerization activity receptor agonist activity low-density lipoprotein particle receptor binding RAGE receptor binding chaperone binding PTB domain binding growth factor receptor binding amylin binding protein serine/threonine kinase binding molecular function activator activity heparan sulfate binding protease inhibitor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter microglial cell activation response to yeast astrocyte activation involved in immune response regulation of transcription from RNA polymerase II promoter regulation of translation cellular copper ion homeostasis endocytosis apoptotic process cell adhesion G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway Notch signaling pathway chemical synaptic transmission axonogenesis central nervous system development learning or memory learning memory mating behavior locomotory behavior axo-dendritic transport negative regulation of cell proliferation associative learning adult locomotory behavior visual learning response to lead ion regulation of gene expression positive regulation of macromolecule metabolic process positive regulation of gene expression negative regulation of gene expression response to auditory stimulus microglia development axon midline choice point recognition neuron remodeling dendrite development calcium-mediated signaling antibacterial humoral response antifungal humoral response regulation of Wnt signaling pathway extracellular matrix organization neuron projection development phospholipase D-activating G-protein coupled receptor signaling pathway positive regulation of chemokine production positive regulation of interferon-gamma production positive regulation of interleukin-1 beta production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production positive regulation of superoxide anion generation regulation of toll-like receptor signaling pathway ionotropic glutamate receptor signaling pathway regulation of multicellular organism growth lipoprotein metabolic process positive regulation of apoptotic process positive regulation of MAPK cascade positive regulation of neuron apoptotic process innate immune response positive regulation of nitric oxide biosynthetic process positive regulation of neuron differentiation positive regulation of G-protein coupled receptor protein signaling pathway positive regulation of Toll signaling pathway positive regulation of glycolytic process positive regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade astrocyte activation regulation of long-term neuronal synaptic plasticity collateral sprouting in absence of injury positive regulation of inflammatory response regulation of synapse structure or activity synapse organization defense response to Gram-negative bacterium defense response to Gram-positive bacterium positive regulation of calcium-mediated signaling positive regulation of cell activation cognition positive chemotaxis positive regulation of membrane protein ectodomain proteolysis regulation of protein metabolic process positive regulation of protein metabolic process protein homooligomerization protein tetramerization regulation of neurotransmitter uptake positive regulation of protein kinase B signaling low-density lipoprotein particle mediated signaling antimicrobial humoral immune response mediated by antimicrobial peptide protein trimerization positive regulation of ERK1 and ERK2 cascade response to interleukin-1 cellular response to copper ion cellular response to manganese ion cellular response to cAMP cellular response to norepinephrine stimulus positive regulation of monocyte chemotaxis negative regulation of canonical Wnt signaling pathway modulation of excitatory postsynaptic potential NMDA selective glutamate receptor signaling pathway positive regulation of gene expression, epigenetic positive regulation of cAMP/PKA signal transduction regulation of spontaneous synaptic transmission negative regulation of protein localization to nucleus regulation of beta-amyloid clearance negative regulation of long-term synaptic potentiation positive regulation of long-term synaptic potentiation positive regulation of NIK/NF-kappaB signaling regulation of calcium ion transmembrane transport via high voltage-gated calcium channel negative regulation of pri-miRNA transcription from RNA polymerase II promoter regulation of dendritic spine maintenance positive regulation of amyloid precursor protein catabolic process regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of blood circulation positive regulation of G protein-coupled receptor internalization positive regulation of endothelin production positive regulation of protein import cellular response to beta-amyloid regulation of presynapse assembly positive regulation of response to endoplasmic reticulum stress regulation of amyloid fibril formation positive regulation of amyloid fibril formation regulation of response to calcium ion amyloid fibril formation cellular response to nerve growth factor stimulus neuron projection maintenance positive regulation of T cell migration positive regulation of excitatory postsynaptic potential positive regulation of extrinsic apoptotic signaling pathway suckling behavior mRNA polyadenylation protein phosphorylation response to oxidative stress regulation of epidermal growth factor-activated receptor activity cholesterol metabolic process negative regulation of endopeptidase activity positive regulation of peptidase activity positive regulation of G2/M transition of mitotic cell cycle forebrain development regulation of protein binding negative regulation of neuron differentiation neuromuscular process controlling balance synaptic growth at neuromuscular junction neuron apoptotic process smooth endoplasmic reticulum calcium ion homeostasis nervous system development negative regulation of peptidase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline