Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
AA Sequence of recombinant protein: DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Betacellulin. Anti-Human Betacellulin-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect hBetacellulin by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Betacellulin (500-P254) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBetacellulin.
Western Blot: To detect hBetacellulin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBetacellulin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P254BT-1MG will be provided as 2 x 500 µg
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Gene Aliases: BTC
UniProt ID: (Human) P35070
Entrez Gene ID: (Human) 685
Molecular Function: growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support