Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • brat Antibodies

          Invitrogen

          brat Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite brat Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          brat Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          brat Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          brat Antibody (PA5-144542) in WB

          Western blot analysis of brat in recombinant protein using a dilution of 1:1,000. Samples were incubated with brat polyclonal antibody (Product # PA5-144542). Secondary: Goat polyclonal to rabbit IgG at 1:50,000 dilution. Predicted band size: 31 kDa. Observed band size: 31 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          brat Antibody in Western Blot (WB)
          brat Polyclonal Antibody

          Product Details

          PA5-144542

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Fruit fly

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant Drosophila melanogaster (Fruit fly) brat protein (294-487aa)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          5 mg/mL

          Amount

          500 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.03% Proclin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3092389

          Target Information

          A NHL-domain family protein that functions a translational repressor to inhibit cell proliferation (PubMed:11274060). Plays a central role in translation repression of hb mRNA by being recruited by nanos (nos) and pum to the Nanos Response Element (NRE), a 16 bp sequence in the hb mRNA 3'-UTR (PubMed:11274060). Probably recruited by other proteins to repress translation of other mRNAs in other tissues (PubMed:11274060). Involved in the regulation of ribosomal RNA synthesis and cell growth (PubMed:11807032). Participates in abdominal segmentation and imaginal disk development (PubMed:11274060). During neuroblast division, segregates asymmetrically and inhibits self-renewal of one of the two daughter cells (PubMed:16564014). Together with the asymmetrically segregating transcription factor prospero ensures that the daughter cell will stop growing, exit the cell cycle, and differentiate into neurons possibly by modulating the function of dm in ganglion mother cells (GMC) (PubMed:11807032, PubMed:16564014). Restricts developmental potential of type II intermediary neuronal progenitor (INP) cells playing a role in proliferation and maturation of the neuroblasts (PubMed:24550111, PubMed:14561773, PubMed:22143802, PubMed:18342578).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Brain tumor protein brat CG10719; brain tumour; brat-PA; brat-PB; brat-PC; brat-PE; brat-PF; CG10719-PA; CG10719-PB; CG10719-PC; CG10719-PE; CG10719-PF; Protein brain tumor

          View more View less

          Gene Aliases: anon-37CDa; BRAT; Cf; CG10719; Dmel\CG10719; Dmel_CG10719; E60; fs(2)ltoPM43; l(2)37Cf; l(2)brat; l(2)E60; lincRNA.S1565

          View more View less

          UniProt ID: (Fruit fly) Q8MQJ9

          View more View less

          Entrez Gene ID: (Fruit fly) 35197

          View more View less

          Function(s)
          mRNA 3'-UTR binding protein binding zinc ion binding translation repressor activity translation regulator activity
          Process(es)
          regulation of translation neuroblast fate determination ganglion mother cell fate determination neuroblast proliferation negative regulation of neuroblast proliferation axon guidance ventral cord development brain development negative regulation of cell proliferation asymmetric cell division regulation of synaptic growth at neuromuscular junction rRNA transcription negative regulation of gene expression neuroblast differentiation neuroblast development negative regulation of translation neuron differentiation segmentation oogenesis regulation of neurogenesis asymmetric neuroblast division resulting in ganglion mother cell formation regulation of synaptic vesicle endocytosis regulation of neuroblast proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline