Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • DGUOK Antibodies

          Proteintech

          dGK Polyclonal Antibody

          View all (4) DGUOK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite dGK Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          dGK Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          dGK Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          dGK Antibody (16459-1-AP) in IP

          IP result of anti-dGK (IP:16459-1-AP, 3ug; Detection:16459-1-AP 1:300) with mouse kidney tissue lysate 4000ug. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          dGK Antibody in Immunoprecipitation (IP)
          dGK Polyclonal Antibody

          Product Details

          16459-1-AP

          Applications
          Tested Dilution
          Publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          dGK Fusion Protein Ag9501 (1-277 aa encoded by BC015757)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Amount

          34.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MAAGRLFLS RLRAPFSSMA KSPLEGVSSS RGLHAGRGPR RLSIEGNIAV GKSTFVKLLT KTYPEWHVAT EPVATWQNIQ AAGTQKACTA QSLGNLLDMM YREPARWSYT FQTFSFLSRL KVQLEPFPEK LLQARKPVQI FERSVYSDRY IFAKNLFENG SLSDIEWHIY QDWHSFLLWE FASRITLHGF IYLQASPQVC LKRLYQRARE EEKGIELAYL EQLHGQHEAW LIHKTTKLHF EALMNIPVLV LDVNDDFSEE VTKQEDLMRE VNTFVKNL (1-277 aa encoded by BC015757)

          Target Information

          Mitochondrial deoxyguanosine kinase (DGUOK) is required for the phosphorylation of several deoxyribonucleosides and certain purine deoxykribonucleoside analogs widely employed as antiviral and chemotherapeutic agents. Purine deoxyribonucleoside analogs are extensively used in treatment of lymphoproliferative disorders. These compounds are administered as pro-drugs, and their efficiency is dependent on intracellular phosphorylation to the corresponding triphosphates. In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predominantly by 2 deoxyribonucleoside kinases: cytosolic deoxycytidine kinase (DCK) and mitochondrial deoxyguanosine kinase (DGUOK also known as DGK). DGUOK expression is ubiquitous, with highest levels in muscle, brain, liver and lymphoid tissues. Defects in DGUOK are a cause of mitochondrial DNA depletion syndrome (MDS). MDS is a clinically heterogeneous group of disorders characterized by a reduction in mitochondrial DNA (mtDNA) copy number. Primary mtDNA depletion is inherited as an autosomal recessive trait and may affect single organs, typically muscle or liver, or multiple tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Deoxyadenosine kinase, mitochondrial; Deoxyguanosine kinase, mitochondrial

          View more View less

          Gene Aliases: DGK; dGK1; dGK2; dGK3; DGUOK; MTDPS3; NCPH; NCPH1; PEOB4

          View more View less

          UniProt ID: (Human) Q16854, (Mouse) Q9QX60

          View more View less

          Entrez Gene ID: (Human) 1716, (Mouse) 27369, (Rat) 297389

          View more View less

          Function(s)
          nucleotide binding deoxyadenosine kinase activity deoxyguanosine kinase activity protein binding ATP binding kinase activity transferase activity deoxynucleoside kinase activity nucleoside kinase activity molecular_function nucleotide kinase
          Process(es)
          nucleobase-containing compound metabolic process guanosine metabolic process negative regulation of neuron projection development mitochondrial ATP synthesis coupled electron transport dGTP metabolic process purine deoxyribonucleoside metabolic process dAMP salvage carbohydrate derivative metabolic process protein phosphorylation deoxyribonucleoside monophosphate biosynthetic process nucleotide biosynthetic process phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline