Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QIVALERMRA ADQTTAEQGM RQLEQENAAL KECRNEYERS LQNHQFELKK LKEEWSQREI VSVAMAQALE EVRKQREEFQ QQAANLTAII DEKEQNLREK TEVLLQKEQE ILQLERGHNS ALLQIHQLQA ELEALRTLKA EEAAVVAEQE DLLRLRGPLQ AEALSVNESH VTSRAMQDPV FQLPTAGRTP NGEVGAMDLT QLQKEKQDLE QQLLEKNKTI KQMQQRMLEL RKTLQKELKI RPDNELFEVR EKPGPEMANM APSVTNNTDL TDAREINFEY LKHVVLKFMS CRESEAFHLI KAVSVLLNFS QEEENMLKET LEYKMSWFGS KPAPKGSIRP SISNPRIPWS
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gap junction protein, alpha 4, 37kD; golgi autoantigen, golgin subfamily a, 1; Golgin subfamily A member 1; Golgin-97; unnamed protein product
Gene Aliases: 0710001G09Rik; 2210418B03Rik; AW107649; GOLGA1; Golgi97; golgin-97
UniProt ID: (Human) Q92805, (Mouse) Q9CW79
Entrez Gene ID: (Human) 2800, (Mouse) 76899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support