Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • nNOS Antibodies

          Invitrogen

          nNOS Polyclonal Antibody

          45 Published Figures
          98 References
          View all (36) nNOS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite nNOS Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (45)
          nNOS Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          nNOS Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 47

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          nNOS Antibody (61-7000) in IHC (F)

          Immunohistochemical staining of frozen rat cerebral cortex using Rb anti-nNOS (Product # 61-7000). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          nNOS Antibody in Immunohistochemistry (Frozen) (IHC (F))
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          nNOS Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          nNOS Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          nNOS Antibody in Immunocytochemistry (ICC/IF)
          nNOS Antibody in Immunocytochemistry (ICC/IF)
          nNOS Antibody in Immunocytochemistry (ICC/IF)
          nNOS Antibody in Immunocytochemistry (ICC/IF)
          nNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          nNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          nNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          nNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          nNOS Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          nNOS Antibody in Immunohistochemistry (Frozen) (IHC (F))
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry, Immunohistochemistry - Free Floating (IHC, IHC (Free))
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry (IHC)
          nNOS Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          nNOS Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot (WB)
          nNOS Antibody in Western Blot, Immunoprecipitation (WB, IP)
          nNOS Antibody in Immunomicroscopy (IM)
          nNOS Antibody in Immunomicroscopy (IM)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          nNOS Polyclonal Antibody

          Product Details

          61-7000

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 16 publications 16 publications

          Immunohistochemistry (IHC)

          -
          View 54 publications 54 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          5-10 µg/mL
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1-2 µg/mL
          View 4 publications 4 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          -
          View 9 publications 9 publications

          ELISA (ELISA)

          0.1-1 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Immunomicroscopy (IM)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A recombinant protein consisting of 195 amino acids from the N-terminus of rat nNOS protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2313734

          Product Specific Information

          This antibody reacts with the ~160 kDa nNOS protein and does not exhibit any cross-reactivity with the related eNOS or iNOS proteins. During development reactivity was confirmed with a ~160 kD band on western blots of rat and mouse brain tissue lysates (20 µg). For use in immunohistochemistry with paraffin embedded tissues some background staining was evident.

          Target Information

          NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: bNOS1; Brain NOS; constitutive NOS; N-NOS; NC-NOS; neuronal nitric oxide synthase; neuronal NOS; nitric oxidase synthase; nitric oxide synthase 1, neuronal; nitric oxide synthase, brain; nNOS; nNOS mu; NOS Type 1; NOS type I; peptidyl-cysteine S-nitrosylase NOS1; Phospho-bNOS; Phospho-Brain NOS; unnamed protein product

          View more View less

          Gene Aliases: 2310005C01Rik; bNOS; N-NOS; NC-NOS; nNOS; NO; NOS; Nos-1; NOS-I

          View more View less

          UniProt ID: (Mouse) Q9Z0J4, (Rat) P29476

          View more View less

          Entrez Gene ID: (Mouse) 18125, (Rat) 24598

          View more View less

          Function(s)
          nitric-oxide synthase activity protein binding calmodulin binding zinc ion binding FMN binding oxidoreductase activity sodium channel regulator activity enzyme binding heme binding peptidyl-cysteine S-nitrosylase activity identical protein binding ion channel binding cadmium ion binding calcium-dependent protein binding flavin adenine dinucleotide binding NADP binding ATPase binding phosphoprotein binding RNA polymerase II sequence-specific DNA binding transcription factor binding NADPH binding scaffold protein binding
          Process(es)
          response to hypoxia regulation of the force of heart contraction regulation of sodium ion transport system process arginine catabolic process nitric oxide biosynthetic process striated muscle contraction cell communication negative regulation of cell proliferation response to heat response to hormone peptidyl-cysteine S-nitrosylation signaling response to lipopolysaccharide multicellular organismal response to stress negative regulation of iron ion transmembrane transport nitric oxide-cGMP-mediated signaling pathway xenobiotic catabolic process vasodilation negative regulation of apoptotic process negative regulation of potassium ion transport response to peptide hormone negative regulation of neuron apoptotic process positive regulation of neuron apoptotic process response to estrogen establishment of protein localization positive regulation of G-protein coupled receptor protein signaling pathway negative regulation of blood pressure positive regulation of transcription, DNA-templated negative regulation of vasoconstriction positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin secretion behavioral response to cocaine regulation of neurogenesis negative regulation of hydrolase activity negative regulation of cytosolic calcium ion concentration negative regulation of serotonin uptake negative regulation of calcium ion transport regulation of postsynaptic membrane potential negative regulation of hepatic stellate cell contraction cellular response to mechanical stimulus cellular response to growth factor stimulus cellular response to epinephrine stimulus positive regulation of adrenergic receptor signaling pathway positive regulation of the force of heart contraction synaptic signaling by nitric oxide positive regulation of adenylate cyclase-activating G protein-coupled receptor signaling pathway positive regulation of long-term synaptic potentiation positive regulation of sodium ion transmembrane transport female pregnancy response to mechanical stimulus response to lead ion response to activity response to nutrient levels response to vitamin E response to vitamin B3 response to nicotine response to decreased oxygen levels regulation of apoptotic process negative regulation of ion transport response to ethanol nitric oxide metabolic process response to nitric oxide regulation of blood circulation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-dg9d8:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0