Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody was generated against full length transgelin 1 protein. It can cross-react with other two transgelins based on the sequence similarity.
Immunogen sequence: MANKGPSYG MSREVQSKIE KKYDEELEER LVEWIIVQCG PDVGRPDRGR LGFQVWLKNG VILSKLVNSL YPDGSKPVKV PENPPSMVFK QMEQVAQFLK AAEDYGVIKT DMFQTVDLFE GKDMAAVQRT LMALGSLAVT KNDGHYRGDP NWFMKKAQEH KREFTESQLQ EGKHVIGLQM GSNRGASQAG MTGYGRPRQI IS (1-201 aa encoded by BC004927)
TAGLN is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 22 kDa actin-binding protein; Actin-associated protein p27; DKFZp686B01212; DKFZp686P11128; epididymis secretory sperm binding protein; Protein WS3-10; SM22; sm22 alpha; SM22-alpha; Sm22alpha; smooth muscle 22 protein; Smooth muscle protein 22-alpha; TAGLN; Transgelin; unnamed protein product
Gene Aliases: SM22; SM22-alpha; Sm22a; SMCC; TAGLN; TAGLN1; TGLN; WS3-10; Ws310
UniProt ID: (Human) Q01995, (Rat) P31232, (Mouse) P37804
Entrez Gene ID: (Human) 6876, (Rat) 25123, (Mouse) 21345
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support