Search
After a solid organ transplant, monitoring levels of donor-derived cell-free DNA (dd-cfDNA) can provide valuable insights into the organ's health and function1. The One Lambda Devyser Accept cfDNA single-tube NGS assay is a minimally invasive solution that helps provide precise and reliable results, to monitor cfDNA following kidney transplantation.
Accept cfDNA is an NGS assay designed to detect and analyze circulating dd-cfDNA in post-transplant human plasma samples. Its unique indel design allows for accurate results, reduces amplification and sequencing errors. Combined with an efficient workflow, Accept cfDNA is the ideal tool for post-transplant monitoring and provides early time to result when run in your own in house HLA lab.
Dual donor testing using One Lambda Devyser Accept cfDNA can support accurately assigning the origin of detected cfDNA from donors in patients with two kidney transplants.
Current testing methods may inaccurately detect elevated levels of cfDNA from the non-functioning graft and can be falsely associated with the second graft in patients with two kidney transplants. These testing methods could be subjected to varying influence from the first graft creating additional complications in the post-transplant monitoring of graft function. The One Lambda Devyser Accept cfDNA test is highly sensitive and has been shown to reliably detect and differentiate cfDNA in patients with two kidney transplants.2
The One Lambda Devyser Accept cfDNA assay was developed by leading transplant specialists to deliver efficiency and ease of use with minimal hands-on time. Labs can receive next-day results from sample to report allowing for timely results. Our dedicated software simplifies data analysis and bioinformatics capabilities, allowing for easy patient tracking in a single location. With the ability to import analyzed data into Histotrac, all information is consolidated, providing clear visibility of trends.
The collaboration between Devyser and Thermo Fisher Scientific combines expertise and resources to offer enhanced and cutting-edge molecular kidney transplant monitoring capabilities to transplant programs, HLA labs, and transplant clinicians. Backed by Thermo Fisher Scientific's robust commercial infrastructure, our collaboration helps support reliable, accurate, and high-quality testing solutions for healthcare providers worldwide.
Product is IVDR certified but not 510(k) cleared. Availability of the product in each country depends on local regulatory marketing authorization status. Dvysr is a trademark of Devyser AB. Not all products are available in all countries. Please consult your local sales representative for details.