Get genes 2 days faster
Invitrogen™ GeneArt™ Gene Synthesis offers the same high-quality genes at the same great price, but now you get a faster turnaround time for genes 1,201–3,000 bp.
|
New turnaround time
(business days)
|
Previous turnaround time
(business days)
|
| Gene length |
Standard | Express | SuperSPEED |
Standard | Express | SuperSPEED |
| < 1,201 bp |
9 | 7 | 5 |
9 | 7 | 5 |
| 1,201–1,800 bp |
12 | 10 | 7 |
14 | 12 | 7 |
| 1,801–3,000 bp |
12 | 10 | NA |
14 | 12 | NA |
Please note: An estimation of total project processing times will be displayed in the cart.
How to order
Just drag and drop the gene synthesis icon, open up the service and fill in all requested information as usual.
Check details >>
GeneArt Strings DNA Libraries
GeneArt™ Strings™ DNA Libraries are linear DNA Fragments from 200 – 2,000 bp, containing up to 3 blocks of degenerate nucleotides. Each block can consist of up to 30 bp using the full IUPAC code. Blocks need to be separated by 30 bp of non-degenerate nucleotides. At least 500 ng are typically shipped within 10-15 business days.
Your advantages
- Affordable: cost-effective alternative to the GeneArt Combinatorial Libraries
- Flexible: full gene design and cloning flexibility; enter your sequence and directly edit, optimize, and order through our online portal
- Fast: typically shipped within 10 - 15 business days
- Comprehensive: the full IUPAC nucleotide code is available (e.g. N, S, K, V, B etc.)
How to order
Drag and Drop the Strings DNA Fragmts & Libs icon, open up the service and activate the Strings DNA Library button. Now you are ready to enter all relevant data.
GeneArt Express Cloning
Using an alternative production procedure with pre-prepared expression vectors, GeneArt™ can clone your synthesized genes directly into selected Invitrogen™ expression plasmids, saving 4-5 business days compared to the standard workflow with cloning into pMx series vector first.
This Express cloning option is different from our subcloning service after gene synthesis. As deliverable, you will receive expression ready genes - your synthesized gene in the selected expression vector, no additional copy in a pMx cloning vector will be provided.
How to order
Just add the express cloning service by clicking the respective box when you fill in your gene synthesis requirements (it’s not necessary to draw a subcloning icon).
We are continually adding new vectors to support your needs. A full list of vectors is available at thermofisher.com/expressgenes
Currently available vectors
- pcDNA3.1(+)
- pcDNA3.3-TOPO
- pcDNA3.4-TOPO
- pFastBac1
- pDONR221
- pET100/D-TOPO
- pET151/D-TOPO
- pRSET A
- pYes2.1V5-His TOPO
Improved Strings DNA fragments functionality
You want to order gene(s) longer than 3,000 bp as Strings™ DNA fragments for assembly to the full length gene(s) in your own lab?
Our newly developed Assembly Assistant can help:
Just use the Strings DNA fragments icon as usual, enter your sequence longer than 3,000 bp and chose the "Assembly Assistant".
The assistant will calculate the fragments < 3,000 bp you can order as Strings and assemble to your full length gene using the GeneArt™ Seamless Cloning and Assembly technology.
GeneArt Strings DNA Fragments can now be ordered from 100 to 3,000 base pairs with improved sequence fidelity.
GeneArt™ Strings™ DNA Fragments are custom-made, uncloned, double-stranded linear DNA fragments from 100 base pairs (bp) to 3,000 bp in length, assembled from synthetic oligonucleotides using the same process developed for GeneArt high-quality gene synthesis. Strings Fragments are delivered dried, ready for resuspension and screening to enable the identification of the correct clone. Strings Fragments are a fast and smart alternative to get your synthetic gene and clone your expression plasmid.
Affordable – Strings DNA Fragments are a cost-effective alternative to gene synthesis
Flexible – full gene design and cloning flexibility; enter your sequence and directly edit, optimize, and order through our online portal
Fast – Strings DNA Fragments – 100 to 3,000 bp – are produced within 5 or 8 business days, ready for cloning and sequencing in your lab.*
More information >>
Project Manager layout and functions
Project Configurator layout
Smarter design – new functions – easier handling
Improved layout and new features allow now for easier set-up and managing of your projects.
1. Project Manager
Improved guidance for starting a project comes along with a new, tabulated view, providing all major project information at a glance. New sorting function and new search function help to find required information. The most recently used project folder will be open by default, project history can be expanded & collapsed.
2. Project Configurator
New design and layout with horizontal grouping of services enables functional displaying of all important information and allows for reduced scrolling and intuitive navigation.
3. Navigation
Guidance through the major ordering steps now is supported by a new navigation bar.
Easy subcloning into your customized vector of choice
Your custom vectors will get available in your individual short list "My portal vectors" as soon as they have been ordered once. Any future cloning into one of these vectors will then require only entering the name plus one click for selection of your vector.