Search
Trizol Reagents are complete, ready-to-use reagents for the isolation of genomic DNA. Genomic DNA isolated with DNAzol Reagents can be used for restriction endonuclease digestion, Southern blot analysis, molecular cloning, and PCR.
The DNAzol Reagent procedure can be completed in typically 10–30 minutes with DNA recoveries of 70–100%. DNAzol Reagent can be used with a broad spectrum of sample materials and allows for rapid isolation and high recovery of genomic DNA (gDNA). Downstream applications include southern blotting, PCR, cloning, restriction endonuclease digestion, and dot blot hybridization.
Learn more about DNAzol Reagent
Learn more about DNAzol BD Reagent
Learn more about Plant DNAzol Reagent
| Sample Type (General) | Final Product | Product Name | Quantity | Cat. No. |
|---|---|---|---|---|
| Cells | Genomic DNA | DNAzol® Reagent | 100 mL | 10503-027 |
| Blood | Genomic DNA | DNAzol® BD Reagent | 100 mL | 10974-020 |
| Plant samples | Genomic DNA | Plant DNAzol® Reagent | 100 mL | 10978-021 |