6420 - Key/Specs/Resources/Doc/Contact
Pore analysis software for Phenom Desktop SEM
Integrated Thermo Scientific Phenom PoroMetric Software allows you to use any Phenom Desktop SEM to gather data on the distribution, size, and aspect ratio of pores in your sample.
Key Features
Easy image acquisition
Acquire images directly from the Phenom Desktop SEM.
Correlate features
Correlate pore features such as area, aspect ratio, as well as major and minor axes.
Improved workflow
Fast and convenient operation improves your workflow and makes scheduling simple and predictable.
Specifications
Style Sheet for Products Table Specifications
Pore size range
  • 100 nm - 0.1 mm
Pore detection
  • 8/16 bit image processing
Speed
  • More than 1,000 pores per minute
Measured properties
  • Size, shape, count
Graphical display
  • Plot graphs of the circle equivalent diameter
  • SEM images and detected pores 
Output
  • Report in docx format
  • TIFF image format
  • CSV file
  • Project file (.POME) for offline analysis
Part of Thermo Scientific Phenom ProSuite Software
  • Network storage enabled
  • Phenom integrated system
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Resources
Contact us
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Electron microscopy support and resources