Structured Illumination Microscopy (SIM)   Structured illumination is used to enhance spatial resolution and involves illuminating the sample with patterned light and using software to analyze the information in Moiré fringes outside the normal range of observation. Reconstruction software deciphers the images at about 2-fold higher resolution than the diffraction limit, or ~100 nm. SIM has advantages over other SRM methods in that it can be used for imaging thicker sections, for 3D imaging, and for live-cell imaging. Image quality increases with bright and photostable dyes as well as precise targeting. Fluorescent proteins are commonly used for SIM investigations of live cells in addition to multiplexing with organic dyes and Qdot® probes.
Live-cell labeling with fluorescent proteins
Live-cell labeling with fluorescent proteins for SIMBecause SIM can be used for thicker sections and live-cell imaging, many publications demonstrate the use of fluorescent proteins for high-resolution studies using GFP or tagRFP. Molecular Probes® CellLight® reagents are ready-to-use fluorescent protein constructs (GFP or tagRFP) targeted to specific subcellular structures. These reagents provide a simple and effective method for introducing targeted intracellular labels within living cells. Specific organelles such as mitochondria, lysosomes, membranes, cytoskeleton, and nucleus can all be visualized using CellLight® reagents in live cells or after fixation. CellLight® reagents can be multiplexed with a range of fluorophores in live cells or fixed and imaged using immunofluorescence techniques.
Antibody conjugates and labels
Antibody conjugates and labels for SIMMolecular Probes® fluorophores have been tried and tested in SIM applications as conjugates of antibodies for fixed-cell applications, and proteins, dextrans, or other biomolecules for fixed or live cells. Use the table below to select the fluorophore with the best wavelength and SIM rating for your application—citations are listed for each. Several Alexa Fluor® probes are outstanding in this application. By following the links, you can also find all of the bioconjugates of each fluorophore currently available to target your molecule of interest; the list ranges from antibodies and phalloidins to growth factors and lectins. If you don't find a conjugate you need on the list, the links will also show you reactive dye forms or optimized labeling kits to enable you to create your own bioconjugate probes.
DNA stains
DNA stains for SIMDAPI is one of the most widely used reagents for DNA staining, and it is highly rated for SIM applications. Because of its high affinity for DNA, it is also frequently used for counting cells, measuring apoptosis, and as a nuclear segmentation tool in high-content imaging analysis. Because DNA is highly abundant, DAPI's limited brightness produces adequate signal and the blue fluorescence is well separated from signals in the FITC channel. Since DAPI is cell-impermeant, it is generally used to stain fixed cells, although the stain will enter live cells when used at higher concentrations.
Organelle stains
Organic dyes are easily multiplexed with fluorescent proteins for either live- or fixed-cell imaging, and some organelle-specific probes are suitable for both. MitoTracker® Red dye is a far-red–fluorescent dye that stains mitochondria in live cells, and its accumulation is dependent upon membrane potential. It serves as an effective SIM probe, and is retained in cells after fixation.
CellMask™ Orange Plasma Membrane Stain allows fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins. This stain is often used as a segmentation tool for HCS (high-content screening) as well as to stain cellular plasma membranes for standard fluorescence microscopy—it also rates very highly for SIM. The stain survives fixation, but not permeabilization, so it is not suitable for experiments that also involve probing internal targets with antibodies.
Mouse melanoma cells
Enlarge
Mouse melanoma cells were labeled with Alexa Fluor® 488 phalloidin (green), MitoTracker® Deep Red FM (magenta), and DAPI (blue) and imaged using SIM. Image courtesy of Catherine Galbraith and James Galbraith, Oregon Health & Science University.
Drosophila
Enlarge
Drosophila S2 cell labeled with Alexa Fluor® 488 phalloidin (blue), anti-tubulin primary antibody and Alexa Fluor® 568 secondary antibody (magenta), and MitoTracker® Deep Red (gold) and imaged using SIM. Image courtesy of William Voss and Catherine Galbraith, Oregon Health & Science University.  
SIM product selection guide
  GFP TagRFP
Target Expressed protein Expressed protein
Bibliography Citations Citations
Laser line (nm) 488 488/532
Standard filter set FITC TRITC
Ex/Em (nm) 488/510 555/584
  Find products Find products
  Alexa Fluor® 405 Alexa Fluor® 488 Alexa Fluor® 546 Alexa Fluor® 555 Alexa Fluor® 568 Alexa Fluor® 594
Target Label Label Label Label Label Label
Bibliography
Citations Citations Citations Citations Citations
Laser line (nm) 350/405 488 488 488 561 594
Standard filter set DAPI FITC TRITC TRITC RFP Texas Red® dye
Ex/Em (nm) 401/421 495/519 556/573 555/580 578/603 590/617
  Find products Find products Find products Find products Find products Find products
  DAPI
Target dsDNA
Bibliography Citations
Laser line (nm) 350/405
Standard filter set DAPI
Ex/Em (nm) 350/470
  Find products
  MitoTracker® Red MitoTracker® Deep Red CellMask™ Orange
Target Mitochondria Mitochondria Cell membrane
Bibliography Citations Citations
Laser line (nm) 561 561 488
Standard filter set Texas Red® dye Cy®5 TRITC
Ex/Em (nm) 581/644 644/665 554/567
Cat. No. M22425 M22426 C10045
  GFP TagRFP
Target Expressed protein Expressed protein
Bibliography Citations Citations
Laser line (nm) 488 488/532
Standard filter set FITC TRITC
Ex/Em (nm) 488/510 555/584
  Find products Find products
  Alexa Fluor® 405 Alexa Fluor® 488 Alexa Fluor® 546 Alexa Fluor® 555 Alexa Fluor® 568 Alexa Fluor® 594
Target Label Label Label Label Label Label
Bibliography
Citations Citations Citations Citations Citations
Laser line (nm) 350/405 488 488 488 561 594
Standard filter set DAPI FITC TRITC TRITC RFP Texas Red® dye
Ex/Em (nm) 401/421 495/519 556/573 555/580 578/603 590/617
  Find products Find products Find products Find products Find products Find products
  DAPI
Target dsDNA
Bibliography Citations
Laser line (nm) 350/405
Standard filter set DAPI
Ex/Em (nm) 350/470
  Find products
  MitoTracker® Red MitoTracker® Deep Red CellMask™ Orange
Target Mitochondria Mitochondria Cell membrane
Bibliography Citations Citations
Laser line (nm) 561 561 488
Standard filter set Texas Red® dye Cy®5 TRITC
Ex/Em (nm) 581/644 644/665 554/567
Cat. No. M22425 M22426 C10045